    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • Vimentin Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Invitrogen
          Vimentin Monoclonal Antibody (SP20)
          Advanced Verification
          4 Published Figures
          39 References
          View all (194) Vimentin antibodies
          Tech Support
          Cite Vimentin Monoclonal Antibody (SP20)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          • Published Figures (4)
          • Advanced Verification (1)
          Vimentin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Vimentin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 8
          Vimentin Antibody (MA5-14564) in ICC/IF
          Immunofluorescent analysis of vimentin in HeLa cells. The cells were fixed with 4% paraformaldehyde in PBS for 15 minutes at room temperature, permeabilized with 0.1% Triton X-100 for 15 minutes and blocked with 3% BSA for 30 minutes at room temperature. Cells were stained with a vimentin rabbit monoclonal antibody (Product # MA5-14564) at a dilution of 1:200 in blocking buffer overnight at 4C, and then incubated with a Goat anti-Rabbit IgG (H+L) Secondary Antibody, Alexa Fluor Plus 488 conjugate (Product # A32731) at a dilution of 1:500 f... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Vimentin Antibody in Immunocytochemistry (ICC/IF)
          Vimentin Antibody in Immunohistochemistry (IHC)
          Vimentin Antibody in Western Blot (WB)
          Vimentin Antibody in Immunocytochemistry (ICC/IF)
          Vimentin Antibody in Immunocytochemistry (ICC/IF)
          Vimentin Antibody in Immunocytochemistry (ICC/IF)
          Vimentin Antibody in Western Blot (WB)
          Vimentin Antibody
          Product Details
          MA5-14564
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:100-1:500
          View 5 publications 5 publications
          Immunohistochemistry (IHC)
          -
          View 23 publications 23 publications
          Immunohistochemistry (Paraffin) (IHC (P))
          1:200
          View 2 publications 2 publications
          Immunohistochemistry (Frozen) (IHC (F))
          1:20
          -
          Immunocytochemistry (ICC/IF)
          1:1,000
          View 8 publications 8 publications
          Flow Cytometry (Flow)
          1:100
          -
          ELISA (ELISA)
          -
          View 1 publication 1 publication
          Product Specifications
          Published species
          Human, Mouse, Non-human primate
          Host/Isotype
          Rabbit / IgG
          Class
          Monoclonal
          Type
          Antibody
          Clone
          SP20
          Immunogen
          Recombinant protein encoding human vimentin
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          Conc. Not Determined
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Wet ice
          RRID
          AB_10981427
          Product Specific Information
          MA5-14564 targets Vimentin in IHC (P) , WB, IF and ICC applications and shows reactivity with Human, Primate, Mouse and Rat samples.
          The MA5-14564 immunogen is recombinant protein encoding human vimentin.
          Target Information
          Vimentin is a developmentally regulated intermediate filament protein (IFP) found in cells of mesenchymal origin. It is believed to be involved with the intracellular transport of proteins between the nucleus and plasma membrane. Unlike other IFP proteins, vimentin is expressed, along with desmin, during the early stages of cellular development. During the development process, vimentin is exchanged for new, tissue-specific IFPs. Vimentin has been implicated to be involved in the rate of steroid synthesis via its role as a storage network for steroidogenic cholesterol containing lipid droplets. Vimentin phosphorylation by a protein kinase causes the breakdown of intermediate filaments and activation of an ATP and myosin light chain dependent contractile event. This results in cytoskeletal changes that facilitate the interaction of the lipid droplets within mitochondria, and subsequent transport of cholesterol to the organelles leading to an increase in steroid synthesis.
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline