    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CDKL2 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Invitrogen
          CDKL2 Monoclonal Antibody (6H8)
          View all (19) CDKL2 antibodies
          Tech Support
          Cite CDKL2 Monoclonal Antibody (6H8)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          CDKL2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CDKL2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CDKL2 Antibody (MA5-20569) in ICC/IF
          Immunofluorescence analysis of HeLa cells using an anti-CDKL2 monoclonal antibody (Product # MA5-20569). (antibody concentration 10 µg/mL). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CDKL2 Antibody in Immunocytochemistry (ICC/IF)
          CDKL2 Monoclonal Antibody (6H8)
          Product Details
          MA5-20569
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1-5 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          10 µg/mL
          -
          ELISA (ELISA)
          Assay-Dependent
          -
          Product Specifications
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          6H8
          Immunogen
          CDKL2 (NP_003939, 394 a.a. ~ 493 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.2-1.0 mg/mL
          Amount
          100 µg
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2606256
          Product Specific Information
          Peptide Sequence: LKDCSNVSVD HTRNPSVAIP PLTHNLSAVA PSINSGMGTE TIPIQGYRVD EKTKKCSIPF VKPNRHSPSG IYNINVTTLV SGPPLSDDSG ADLPQMEHQH
          Target Information
          The activation of signal transduction pathways by growth factors, hormones and neurotransmitters is mediated by the MAP kinases ERK 1 and ERK 2. ERK proteins are regulated by dual phosphorylation at specific tyrosine and threonine sites mapping within a characteristic Thr-Glu-Tyr motif. The protein kinase p56 KKIAMRE is distantly related to the MAP kinase group of proteins and is closely related to p42 KKIALRE. KKIAMRE is predominantly expressed in testis, kidney, brain and lung. KKIAMRE contains the conserved MAP kinase dual phosphorylation motif in the sequence Thr-Asp-Tyr and is activated by treatment of cells by EGF. However, unlike other MAP kinases, the EGF-stimulated kinase activity does not require phosphorylation of KKIAMRE and KKIALRE in the Thr-Asp-Tyr motif.
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline