    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO and CRO Solutions
          • Primary Antibodies ›
          • ARMC6 Antibodies
          Invitrogen
          ARMC6 Monoclonal Antibody (2H8)
          View all (9) ARMC6 antibodies
          Tech Support
          Cite ARMC6 Monoclonal Antibody (2H8)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          ARMC6 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ARMC6 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          ARMC6 Antibody (MA5-22880) in WB
          Western Blot analysis of ARMC6 expressed in transfected 293T cell line by anti-ARMC6 monoclonal antibody (Product # MA5-22880), clone 2H8. Lane 1: ARMC6 transfected lysate (Predicted MW: 51.5 KDa). Lane 2: Non-transfected lysate. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ARMC6 Antibody in Western Blot (WB)
          ARMC6 Antibody in Western Blot (WB)
          ARMC6 Antibody in Immunoprecipitation (IP)
          ARMC6 Antibody in ELISA (ELISA)
          ARMC6 Monoclonal Antibody (2H8)
          Product Details
          MA5-22880
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1-5 µg/mL
          -
          ELISA (ELISA)
          Assay-Dependent
          -
          Immunoprecipitation (IP)
          Assay-Dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2H8
          Immunogen
          ARMC6 (AAH67125, 1 a.a. ~ 476 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1.0 mg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.4
          Contains
          no preservative
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2607264
          Product Specific Information
          Peptide Sequence: MVSKRIAQET FDAAVRENIE EFAMGPEEAV KEAVEQFESQ GVDLSNIVKT APKVSADGSQ EPTHDILQML SDLQESVASS RPQEVSAYLT RFCDQCKQDK ACRFLAAQKG AYPIIFTAWK LATAGDQGLL LQSLNALSVL TDGQPDLLDA QGLQLLVATL TQNADEADLT CSGIRCVRHA CLKHEQNRQD LVKAGVLPLL TGAITHHGHH TDVVREACWA LRVMTFDDDI RVPFGHAHNH AKMIVQENKG LKVLIEATKA FLDNPGILSE LCGTLSRLAI RNEFCQEVVD LGGLSILVSL LADCNDHQMR DQSGVQELVK QVLSTLRAIA GNDDVKDAIV RAGGTESIVA AMTQHLTSPQ VCEQSCAALC FLALRKPDNS RIIVEGGGAV AALQAMKAHP QKAGVQKQAC MLIRNLVAHG QAFSKPILDL GAEALIMQAR SAHRDCEDVA KAALRDLGCH VELRELWTGQ RGNLAP
          Target Information
          The function of this gene's protein product has not been determined. A related protein in mouse suggests that this protein has a conserved function.
          Bioinformatics
          Protein Aliases: Armadillo repeat-containing protein 6; unnamed protein product
          View more View less
          Gene Aliases: ARMC6; R30923_1
          View more View less
          UniProt ID: (Human) Q6NXE6
          View more View less
          Entrez Gene ID: (Human) 93436
          View more View less
          Process(es)
          hematopoietic progenitor cell differentiation
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline