    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • EMP Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Invitrogen
          EMP Monoclonal Antibody (730340)
          View all (6) EMP antibodies
          Tech Support
          Cite EMP Monoclonal Antibody (730340)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          EMP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          EMP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          EMP Antibody (MA5-24344) in ICC/IF
          Immunocytochemical analysis of EMP/MAEA was detected in immersion fixed Jurkat human acute T cell leukemia cell line using mouse Anti-human/mouse/Rat EMP/MAEA Monoclonal Antibody (Product # MA5-24344) at 10 µg/mL for 3 hours at room temperature. Cells were stained using the 557-conjugated Anti-mouse IgG Secondary Antibody (red, upper pane and counterstained with DAPI (blue, lower panel). Specific staining was localized to cell surfaces and cytoplasm. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          EMP Antibody in Immunocytochemistry (ICC/IF)
          EMP Antibody in Western Blot (WB)
          EMP Monoclonal Antibody (730340)
          Product Details
          MA5-24344
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.5 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          8-25 µg/mL
          -
          Product Specifications
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          730340
          Immunogen
          E. coli-derived recombinant human EMP/MAEA Met12-Ser66
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Wet ice
          RRID
          AB_2607102
          Product Specific Information
          Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
          Target Information
          This gene product mediates the attachment of erythroblasts to macrophages. This attachment promotes terminal maturation and enucleation of erythroblasts, presumably by suppressing apoptosis. This protein is an integral membrane protein with the N-terminus on the extracellular side and the C-terminus on the cytoplasmic side of the cell. Two immunologically related isoforms of erythroblast macrophage protein with apparent molecular weights of 33 kD and 36 kD were detected in macrophage membranes, this gene encodes the larger isoform. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity of some variants has not been determined.
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline