    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • FNDC8 Antibodies
          Invitrogen
          FNDC8 Monoclonal Antibody (OTI1F4)
          View all (13) FNDC8 antibodies
          Tech Support
          Cite FNDC8 Monoclonal Antibody (OTI1F4)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          FNDC8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FNDC8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          FNDC8 Antibody (MA5-26331) in IHC (P)
          Immunohistochemistry was performed on paraffin-embedded human lymph node tissue. To expose target proteins, 1 mM EDTA in 10mM Tris, pH8.5, 120°C for 3min was used. Following antigen retrieval, tissues were probed with a FNDC8 monoclonal antibody (Product # MA5-26331). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          FNDC8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FNDC8 Antibody in Western Blot (WB)
          FNDC8 Monoclonal Antibody (OTI1F4)
          Product Details
          MA5-26331
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:4,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:150
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI1F4
          Immunogen
          Full length human recombinant protein of FNDC8 produced in HEK293T cell
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Affinity Chromatography
          Storage buffer
          PBS, pH 7.3, with 1% BSA, 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2724312
          Target Information
          FNDC8 is a protein coding gene.
          Bioinformatics
          Protein Aliases: Fibronectin type III domain-containing protein 8; unnamed protein product
          View more View less
          Gene Aliases: FNDC8
          View more View less
          UniProt ID: (Human) Q8TC99
          View more View less
          Entrez Gene ID: (Human) 54752
          View more View less
          Function(s)
          protein binding
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline