  • Applications
    • Bioprocesamiento
    • Cultivo celular y transfección
    • Terapia celular y génica
    • Cromatografía
    • Pruebas moleculares
    • Soluciones digitales
    • Extracción y análisis de ADN y ARN
    • Espectroscopía, análisis elemental y de isótopos
    • Ver todas las aplicaciones y técnicas
  • Servicios
    • Servicios 360° de CDMO y CRO
    • Servicios de CDMO
    • Servicios de CRO
    • Servicios personalizados
    • Servicios de leasing y financiación
    • Servicios de instrumentos
    • Informática de laboratorio
    • OEM y oferta comercial
    • Servicios de formación
    • Unity Lab Services
    • Ver todos los servicios
  • Ayuda y soporte técnico
    • Regístrese para obtener una cuenta
    • Cómo hacer un pedido
    • Asistencia para el instrumental
    • Centros de soporte técnico
    • Centros de formación
    • Vea todos los temas de ayuda y soporte técnico
      Cell Culture Plastics
  • A quiénes brindamos nuestros servicios
    • Industria biotecnológica
    • Sector biofarmacéutico
    • CDMO
    • Diagnósticos de laboratorio
    • Ciencias aplicadas e industriales
  • Promociones
  • Contacto
  • Pedido rápido
  • Estado del pedido y seguimiento
  • Documentos y certificados
Thermo Fisher Scientific Logo
Search
Buscar
Search button
          • Estado del pedido
          • Pedido rápido
            • Cuenta
            • Pedidos
            • Connect: laboratorio, datos, aplicaciones
            • Productos y proyectos personalizados
            • Services Central
          • Primary Antibodies ›
          • CD45 Antibodies
          Invitrogen
          CD45 Monoclonal Antibody (IH-1)
          View all (459) CD45 antibodies
          Tech Support
          Cite CD45 Monoclonal Antibody (IH-1)
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          CD45 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD45 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CD45 Antibody (MA5-28390) in Flow
          Flow cytometry of CD45 in guinea pig bone marrow. Samples were incubated with CD45 monoclonal antibody (Product # MA5-28390). Image details are as follows: A) BM-MSCs were harvested and cultured from guinea pigs’ bone marrow from healthy (sham-exposed) or CS-exposed individuals. B) Cell characterization was determined by plastic adhesion, colony forming capacity, and morphology. C) Purity was checked by flow cytometric analysis of CD45 expression in BM-MSCs and compared with whole blood. Fluorescence intensity of the no-antibody (left pane... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD45 Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          MA5-28390
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Flow Cytometry (Flow)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Guinea pig
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          IH-1
          Immunogen
          Guinea pig leukemic B lymphocyte L2C cells
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FITC
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          200 µg
          Purification
          Protein A
          Storage buffer
          TBS
          Contains
          <0.1% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2745353
          Target Information
          CD45 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase (PTP) ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45 is absent on non-hematopoietic cell lines, normal and malignant, non-hematopoietic tissues. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists as multiple isoforms as a result of alternative splicing, differ in their extracellular domains but share identical transmembrane and cytoplasmic domains. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells. CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. CD45 has been shown to be an essential regulator of T- and B-cell antigen receptor signaling and suppresses JAK kinases to regulate cytokine receptor signaling. CD45 is also important in promoting cell survival by modulating integrin-mediated signal transduction pathway, DNA fragmentation during apoptosis and inhibition or upregulation of various immunological functions.
          Bioinformatics
          Protein Aliases: T220 and B220
          View more View less
          Entrez Gene ID: (Guinea pig) 100730564
          View more View less
          Function(s)
          protein phosphatase protein modifying enzyme
          Pedidos Plus Icon Minus Icon
          • Estado del pedido
          • Ayuda con el pedido
          • Pedido rápido
          • Centro de suministros
          • eProcurement (B2B)
          Asistencia Plus Icon Minus Icon
          • Ayuda y soporte técnico
          • Contacto
          • Centros de soporte técnico
          • Documentos y certificados
          • Informar de un problema del sitio web
          Recursos Plus Icon Minus Icon
          • Centros de formación
          • Promociones
          • Eventos y seminarios web
          • Redes sociales
          Acerca de Thermo Fisher Plus Icon Minus Icon
          • Acerca de nosotros Acerca de nosotros
          • Carreras profesionales Carreras profesionales
          • Inversores Inversores
          • Noticias Noticias
          • Responsabilidad social Responsabilidad social
          • Marcas registradas
          • Políticas y avisos
          Nuestra cartera Plus Icon Minus Icon
          Spain flag icon
          Spain
          Your items have has been added!
          Host server : magellan-search-blue-74d7b4b4d9-p95z8:80/100.66.74.151:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline