    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • CD4 Antibodies
          Invitrogen
          CD4 Monoclonal Antibody (3-4F4), FITC
          View all (607) CD4 antibodies
          Tech Support
          Cite CD4 Monoclonal Antibody (3-4F4), FITC
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          CD4 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD4 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CD4 Antibody (MA5-28776) in Flow
          Flow cytometry analysis of CD4 in feline peripheral blood lymphocytes. Cells were stained using CD4 Monoclonal Antibody (3-4F4), FITC (Product # MA5-28776) and Mouse Anti-Feline CD8-PE. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD4 Antibody in Flow Cytometry (Flow)
          Product Details
          MA5-28776
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          1 µg/10^6 cells
          -
          Product Specifications
          Species Reactivity
          Cat
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3-4F4
          Immunogen
          Feline T cells
          Conjugate
          FITC FITC FITC
          • Unconjugated
          • Biotin 
          • PE 
          • Request custom conjugation
          Excitation/Emission Max
          498/517 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          500 µg
          Purification
          purified
          Storage buffer
          PBS, pH 7.4
          Contains
          <0.1% sodium azide
          Storage conditions
          4°C, store in dark
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2745735
          Target Information
          The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: CD4 antigen; CD4 antigen p55; cd4a; fCD4; Leu-3; T-cell surface glycoprotein CD4
          View more View less
          Entrez Gene ID: (Cat) 493775
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline