    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
          • Order Status
          • 快速訂購
          • 登入
            登入
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • HPV L1 Capsid Antibodies
          Invitrogen
          HPV pan-specific L1 Monoclonal Antibody (5307)
          View all (2) HPV L1 Capsid antibodies
          Tech Support
          Cite HPV pan-specific L1 Monoclonal Antibody (5307)
          Additional Information:
          {{banner.description}}
          Product Details
          MA5-33266
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-Dependent
          -
          Immunohistochemistry (IHC)
          Assay-Dependent
          -
          ELISA (ELISA)
          1:20-1:200
          -
          Product Specifications
          Species Reactivity
          Virus
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          5307
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          100 µg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.2
          Contains
          0.1% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2815404
          Product Specific Information
          Reactive with the L1 of types 6, 11, 16, 18, 31, 33, 45, 52, & 58
          Target Information
          The papillomavirus major capsid protein, L1, is a ~55 kD protein with the ability to spontaneously self-assemble into virus-like particles (VLPs). These VLPs present an exterior surface essentially indistinguishable from the native 60 nm non-enveloped papillomavirus virion. Purified recombinant L1 proteins can achieve this complex assembly reaction in the absence of any chaperones (Schiller and Lowy, 2012). PMID: 23800545.
          Bioinformatics
          Protein Aliases: Human Papillomavirus; Human Papillomavirus L1; Human Papillomavirus L1 Pan
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline