Specificity: Human CHL1/LICAM2/CALL
The protein encoded by this gene is a member of the L1 gene family of neural cell adhesion molecules. It is a neural recognition molecule that may be involved in signal transduction pathways. The deletion of one copy of this gene may be responsible for mental defects in patients with 3p- syndrome. Several alternatively spliced transcript variants of this gene have been described, but their full length nature is not known.
Protein Aliases: cell adhesion molecule with homology to L1CAM (close homolog of L1); cell adhesion molecule with homology to L1CAM (close homologue of L1); Close homolog of L1; L1 cell adhesion molecule 2; member of neural CAM superfamily; Neural cell adhesion molecule L1-like protein; unnamed protein product
Gene Aliases: CALL; CHL1; L1CAM2
UniProt ID: (Human) O00533
Entrez Gene ID: (Human) 10752