    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
          • Primary Antibodies ›
          • Carbonic Anhydrase IV Antibodies
          Invitrogen
          Carbonic Anhydrase IV Monoclonal Antibody (C16)
          View all (17) Carbonic Anhydrase IV antibodies
          Tech Support
          Cite Carbonic Anhydrase IV Monoclonal Antibody (C16)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          Carbonic Anhydrase IV Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Carbonic Anhydrase IV Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          Carbonic Anhydrase IV Antibody (MA5-43925) in ICC/IF
          Immunocytochemistry analysis of Carbonic Anhydrase IV in Hela cells using Carbonic Anhydrase IV Monoclonal Antibody (C16) (Product # MA5-41559) at a dilution of 30 µg/mL, followed by secondary antibody staining with 2 µg/mL AF488-Linked Caprine Anti-Mouse IgG Polyclonal Antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Carbonic Anhydrase IV Antibody in Immunocytochemistry (ICC/IF)
          Carbonic Anhydrase IV Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Carbonic Anhydrase IV Antibody in Western Blot (WB)
          Carbonic Anhydrase IV Antibody in Western Blot (WB)
          Carbonic Anhydrase IV Monoclonal Antibody (C16)
          Product Details
          MA5-43925
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.01-2 µg/mL
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          5-20 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          5-30 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Mouse / IgG2b, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          C16
          Immunogen
          Recombinant protein Carbonic Anhydrase IV. The antigen corresponds to amino acid range 19-283 of the target protein.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          20 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2912857
          Target Information
          Carbonic anhydrases are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. This gene encodes a glycosylphosphatidyl-inositol-anchored membrane isozyme expressed on the luminal surfaces of pulmonary capillaries and proximal renal tubules. Its exact function is not known, however, it may have a role in inherited renal abnormalities of bicarbonate transport.
          Bioinformatics
          Protein Aliases: CA-IV; CA4; Carbonate dehydratase IV; Carbonic anhydrase 4; Carbonic anhydrase IV; carbonic dehydratase IV; unnamed protein product
          View more View less
          Gene Aliases: CA4; CAIV; Car4; RP17
          View more View less
          UniProt ID: (Human) P22748, (Rat) P48284
          View more View less
          Entrez Gene ID: (Human) 762, (Rat) 29242
          View more View less
          Function(s)
          carbonate dehydratase activity protein binding zinc ion binding lyase activity metal ion binding
          Process(es)
          bicarbonate transport one-carbon metabolic process response to drug animal organ development response to steroid hormone
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          India flag icon
          India
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline