    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • Clostridium difficile GDH Antibodies
          Invitrogen
          Clostridium difficile GDH Monoclonal Antibody (5196)
          View all (5) Clostridium difficile GDH antibodies
          Tech Support
          Cite Clostridium difficile GDH Monoclonal Antibody (5196)
          Additional Information:
          {{banner.description}}
          Product Details
          MA5-47561
          Applications
          Tested Dilution
          Publications
          ELISA (ELISA)
          1:20-1:200
          -
          Product Specifications
          Species Reactivity
          Bacteria
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          5196
          Immunogen
          Proprietary sequence
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          100 µg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.2
          Contains
          0.1% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2942542
          Product Specific Information
          Specificity: Specific for glutamate dehydrogenase of C. difficile. Non-reactive with mammalian cells.
          Best ELISA pairs (capture/conjugate): Clostridium difficile GDH Monoclonal antibody (Product # MA5-47557)/Clostridium difficile GDH Monoclonal antibody (Product # MA5-47561); Clostridium difficile GDH Monoclonal antibody (Product # MA5-47558)/Clostridium difficile GDH Monoclonal antibody (Product # MA5-47561); Clostridium difficile GDH Monoclonal antibody (Product # MA5-47559)/Clostridium difficile GDH Monoclonal antibody (Product # MA5-47561); Clostridium difficile GDH Monoclonal antibody (Product # MA5-47560)/Clostridium difficile GDH Monoclonal antibody (Product # MA5-47561).
          Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 µg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
          Target Information
          Clostridium difficile produces an NAD-specific glutamate dehydrogenase (GDH), which converts l-glutamate into alpha-ketoglutarate through an irreversible reaction. The enzyme GDH is detected in the stool samples of patients with C. difficile-associated disease and serves as one of the diagnostic tools to detect C. difficile infection (CDI). [PMID: 24145018]
          Bioinformatics
          Protein Aliases: GDH
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline