    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GSTM1 Antibodies
          Invitrogen
          GSTM1 Recombinant Rabbit Monoclonal Antibody (23GB4940)
          View all (22) GSTM1 antibodies
          Tech Support
          Cite GSTM1 Recombinant Rabbit Monoclonal Antibody (23GB4940)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          GSTM1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          GSTM1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          GSTM1 Antibody (MA5-52765) in ICC/IF
          Immunocytochemistry analysis of GSTM1 in HepG2 cells. Sample was incubated with GSTM1 monoclonal (Product # MA5-52764) antibody at a dilution of 1:1,000, followed by DAPI and Alexa Fluor 647. Protein abundance based on laser intensity and smart gain: Medium. Scale bar: 20 μm. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GSTM1 Antibody in Immunocytochemistry (ICC/IF)
          GSTM1 Antibody in Western Blot (WB)
          GSTM1 Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          GSTM1 Recombinant Rabbit Monoclonal Antibody (23GB4940)
          Product Details
          MA5-52765
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:5,000
          -
          Immunocytochemistry (ICC/IF)
          1:100-1:1,000
          -
          Flow Cytometry (Flow)
          1:2,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          23GB4940
          Immunogen
          A synthesized peptide derived from human Glutathione S Transferase mu (100-152AA).
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3249242
          Target Information
          Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-transferase that belongs to the mu class. The mu class of enzymes functions in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding the mu class of enzymes are organized in a gene cluster on chromosome 1p13. 3 and are known to be highly polymorphic. These genetic variations can change an individual's susceptibility to carcinogens and toxins as well as affect the toxicity and efficacy of certain drugs. Null mutations of this class mu gene have been linked with an increase in a number of cancers, likely due to an increased susceptibility to environmental toxins and carcinogens. Multiple protein isoforms are encoded by transcript variants of this gene.
          Bioinformatics
          Protein Aliases: Glutathione S-transferase GT8.7; glutathione S-transferase M1; Glutathione S-transferase Mu 1; Glutathione S-transferase Yb-1; glutathione S-transferase Yb-1 subunit; glutathione-S-transferase mu type 1 (Yb1); glutathione-S-transferase, mu 1; glutathione-S-transferase, mu type 1 (Yb1); GST 1-1; GST 3-3; GST class-mu 1; GST HB subunit 4; GST Yb1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GTH4; pmGT10
          View more View less
          Gene Aliases: GST1; GSTA3; Gstb-1; Gstb1; GSTM1
          View more View less
          UniProt ID: (Mouse) Q58ET5, (Rat) P04905
          View more View less
          Entrez Gene ID: (Mouse) 14862, (Rat) 24423
          View more View less
          Function(s)
          glutathione transferase activity steroid binding protein binding nickel cation binding transferase activity protein homodimerization activity protein heterodimerization activity enzyme binding glutathione binding transferase
          Process(es)
          glutathione metabolic process metabolic process cellular response to drug sensory perception of smell response to metal ion response to lithium ion response to lead ion nitrobenzene metabolic process xenobiotic catabolic process response to drug response to amino acid response to ethanol response to antibiotic response to axon injury cellular detoxification of nitrogen compound
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-green-dd86746ff-hszvs:80/100.66.78.131:80.
          git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.53.1-Offline