    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FDPS Antibodies
          Invitrogen
          FDPS Recombinant Rabbit Monoclonal Antibody (23GB1395)
          Advanced Verification
          View all (28) FDPS antibodies
          Tech Support
          Cite FDPS Recombinant Rabbit Monoclonal Antibody (23GB1395)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          • Advanced Verification (1)
          FDPS Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FDPS Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          FDPS Antibody (MA5-53979) in ICC/IF
          Immunocytochemistry analysis of FDPS in HepG2 cells. Nuclei were stained blue with DAPI; FDPS recombinant monoclonal antibody (Product # MA5-53978) with a dilution of 1:1,000 was stained magenta with Alexa Fluor 647. Images were taken using Leica stellaris 5. Protein abundance based on laser Intensity and smart gain: Medium. Scale bar: 20 μm. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          FDPS Antibody in Immunocytochemistry (ICC/IF)
          FDPS Antibody in Western Blot (WB)
          FDPS Antibody in Flow Cytometry (Flow)
          FDPS Antibody
          FDPS Recombinant Rabbit Monoclonal Antibody (23GB1395)
          Product Details
          MA5-53979
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:5,000
          -
          Immunocytochemistry (ICC/IF)
          1:100-1:1,000
          -
          Flow Cytometry (Flow)
          1:200-1:2,000
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          23GB1395
          Immunogen
          A synthesized peptide derived from human FDPS (370-410AA).
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.6 mg/mL
          Amount
          12 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3248450
          Target Information
          The isoprene biosynthetic pathway supply the cell with cholesterol, ubiquinone, and various nonsterol metabolites. The farnesylpyrophosphate synthetase enzyme catalyzes the formation of geranyl and farnesylpyrophosphate from isopentenylpyrophosphate and dimethylallyl pyrophosphate. Analysis of FDPS activity and protein in rat liver, accompanied by immunofluorescence and immunoelectron microscopy studies, demonstrated that FDPS is predominantly localized in peroxisomes. 1 Liver tissue from patients with the peroxisomal deficiency diseases Zellweger syndrome and neonatal adrenoleukodystrophy exhibit diminished activities of FDPS and subsequent isoprenoid synthesis.
          Bioinformatics
          Protein Aliases: (2E,6E)-farnesyl diphosphate synthase; Dimethylallyltranstransferase; Farnesyl diphosphate synthase; Farnesyl pyrophosphate synthase; farnesyl pyrophosphate synthetase; farnesyl pyrophosphate synthetase (EC 2.5.1.1); farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, geranyltranstransferase; FPP synthase; FPP synthetase; Geranyltranstransferase; KIAA1293; unnamed protein product
          View more View less
          Gene Aliases: FDPS; FPPS; FPS; KIAA1293; POROK9
          View more View less
          UniProt ID: (Human) P14324
          View more View less
          Entrez Gene ID: (Human) 2224
          View more View less
          Function(s)
          RNA binding dimethylallyltranstransferase activity geranyltranstransferase activity prenyltransferase activity protein binding transferase activity transferase activity, transferring alkyl or aryl (other than methyl) groups metal ion binding
          Process(es)
          lipid metabolic process steroid biosynthetic process cholesterol biosynthetic process steroid metabolic process cholesterol metabolic process isoprenoid biosynthetic process sterol biosynthetic process geranyl diphosphate biosynthetic process farnesyl diphosphate biosynthetic process
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline