    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • GBP6 Antibodies
          Invitrogen
          GBP6 Monoclonal Antibody (K92016_8F12)
          View all (3) GBP6 antibodies
          Tech Support
          Cite GBP6 Monoclonal Antibody (K92016_8F12)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          GBP6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          GBP6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          GBP6 Antibody (MA5-56357) in IHC (P)
          Immunohistochemistry analysis of GBP6 in paraffin-embedded esophagus tissue. Sample was incubated with GBP6 monoclonal antibody (Product # MA5-55977) at a dilution of 10 µg/mL (RT, 1 hour). Antigen was retrieved through addition of boiling Tris/EDTA buffer pH 9 in a pressure cooker for 3 min. Endogenous peroxidase activity was quenched by incubating the sections with 3% H2O2 for 30 min at room temperature. ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GBP6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GBP6 Antibody in Western Blot (WB)
          GBP6 Antibody in Immunoprecipitation (IP)
          GBP6 Monoclonal Antibody (K92016_8F12)
          Product Details
          MA5-56357
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:100
          -
          Immunoprecipitation (IP)
          1:100
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          K92016_8F12
          Immunogen
          Recombinant human GBP6 (immunogen range is 291-570).
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          200 µg
          Purification
          Protein A
          Storage buffer
          PBS with 50% glycerol, 0.5% BSA
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          RRID
          AB_3668589
          Product Specific Information
          Sequence of this protein is as follows: RLGTLAVTYV EAINSGAVPC LENAVITLAQ RENSAAVQRA ADYYSQQMAQ RVKLPTDTLQ ELLDMHAACE REAIAIFMEH SFKDENQEFQ KKFMETTMNK KGDFLLQNEE SSVQYCQAKL NELSKGLMES ISAGSFSVPG GHKLYMETKE RIEQDYWQVP RKGVKAKEVF QRFLESQMVI EESILQSDKA LTDREKAVAV DRAKKEAAEK EQELLKQKLQ EQQQQMEAQD KSRKENIAQL KEKLQMEREH LLREQIMMLE HTQKVQNDWL HEGFKKKYEE
          Target Information
          Binds GTP, GDP and GMP.
          Bioinformatics
          Protein Aliases: GBP-6; GTP-binding protein 6; Guanine nucleotide-binding protein 6; Guanylate-binding protein 6; unnamed protein product
          View more View less
          Gene Aliases: GBP6
          View more View less
          UniProt ID: (Human) Q6ZN66
          View more View less
          Entrez Gene ID: (Human) 163351
          View more View less
          Function(s)
          nucleotide binding GTPase activity protein binding GTP binding hydrolase activity
          Process(es)
          immune system process immune response defense response to bacterium defense response to protozoan innate immune response defense response to Gram-positive bacterium cellular response to interferon-gamma
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline