    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • SEC24A Antibodies
          Invitrogen
          SEC24A Recombinant Rabbit Monoclonal Antibody (PSH04-36)
          View all (8) SEC24A antibodies
          Tech Support
          Cite SEC24A Recombinant Rabbit Monoclonal Antibody (PSH04-36)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (7)
          SEC24A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SEC24A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 7
          SEC24A Antibody (MA5-56416) in ICC/IF
          Immunocytochemistry analysis of SEC24A with Caco-2 cells. Cells were fixed in 4% paraformaldehyde (20 min, room temp), permeabilized with 0.1% Triton X-100 in PBS (5 min, room temp), then blocked with 1% BSA in 10% negative goat serum (1 hr, room temp). Next they were incubated with SEC24A Recombinant Monoclonal Antibody (Product # MA5-56416)at a dilution of 1:100 in 1% BSA in PBST overnight at 4 &(8)451;. Followed by Goat Anti-Rabbit IgG H&L at a dilution of 1:1,000. DAPI was used to stain the cell nuclei (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SEC24A Antibody in Immunocytochemistry (ICC/IF)
          SEC24A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC24A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC24A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC24A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC24A Antibody in Western Blot (WB)
          SEC24A Antibody in Flow Cytometry (Flow)
          SEC24A Recombinant Rabbit Monoclonal Antibody (PSH04-36)
          Product Details
          MA5-56416
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:200-1:1,000
          -
          Immunocytochemistry (ICC/IF)
          1:100
          -
          Flow Cytometry (Flow)
          1:1,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Non-human primate, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          PSH04-36
          Immunogen
          Recombinant protein within human SEC24A aa 1-350/1,093.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.4, with 0.1% BSA, 40% glycerol
          Contains
          0.05% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3679471
          Target Information
          SC24A is a component of coat protein II (COPII; see MIM 601924)-coated vesicles that mediate protein transport from the endoplasmic reticulum. SEC24A is 1 of several mammalian proteins that show structural and functional homology to yeast Sec24.
          Bioinformatics
          Protein Aliases: Protein transport protein Sec24A; SEC24 family, member A; SEC24 related gene family, member A; SEC24-related protein A; unnamed protein product
          View more View less
          Gene Aliases: SEC24A
          View more View less
          UniProt ID: (Human) O95486, (Mouse) Q3U2P1
          View more View less
          Entrez Gene ID: (Human) 10802, (Mouse) 77371, (Rat) 287275
          View more View less
          Function(s)
          SNARE binding protein binding zinc ion binding metal ion binding molecular_function vesicle coat protein
          Process(es)
          intracellular protein transport ER to Golgi vesicle-mediated transport protein transport vesicle-mediated transport regulation of cholesterol transport cholesterol homeostasis positive regulation of protein secretion cargo loading into COPII-coated vesicle transport regulation of low-density lipoprotein particle receptor biosynthetic process positive regulation of cholesterol homeostasis
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline