FIGURE: 1 / 8
This gene encodes a transcription factor which coactivates transcription of estrogen receptor responsive genes and corepresses genes activated by other hormone receptors or sequence-specific transcription factors. Expression of this gene is regulated by both members of the estrogen receptor family. This gene may be involved in the progression of several types of cancer. Alternative splicing results in multiple transcript variants.
Protein Aliases: Modulator of non-genomic activity of estrogen receptor; modulator of nongenomic activity of estrogen receptor; proline and glutamic acid rich nuclear protein; proline, glutamic acid and leucine rich protein 1; Proline-, glutamic acid- and leucine-rich protein 1; proline-, glutamic acid-, leucine-rich protein 1; scaffold protein; Transcription factor HMX3; unnamed protein product
Gene Aliases: 4930563C04Rik; HMX3; MNAR; P160; PELP1; RGD1306320
UniProt ID: (Human) Q8IZL8, (Rat) Q56B11, (Mouse) Q9DBD5
Entrez Gene ID: (Human) 27043, (Rat) 360552, (Mouse) 75273