          • Primary Antibodies ›
          • ATE1 Antibodies
          Invitrogen
          ATE1 Recombinant Rabbit Monoclonal Antibody (24GB4720)
          View all (13) ATE1 antibodies
          Tech Support
          Cite ATE1 Recombinant Rabbit Monoclonal Antibody (24GB4720)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          ATE1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ATE1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          ATE1 Antibody (MA5-57344) in ICC/IF
          Immunocytochemical staining of ATE1 in HepG2 cells using ATE1 Recombinant Monoclonal Antibody (Product # MA5-57343) at a 1:1,1000 dilution. Nuclei were stained blue with DAPI; Arginyltransferase 1 was stained magenta with Alexa Fluor® 647. Images were taken using Leica stellaris 5. Protein abundance based on laser Intensity and smart gain: Low. Scale bar: 20 μm. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ATE1 Antibody in Immunocytochemistry (ICC/IF)
          ATE1 Antibody in Western Blot (WB)
          ATE1 Antibody in Western Blot (WB)
          ATE1 Antibody in Flow Cytometry (Flow)
          ATE1 Recombinant Rabbit Monoclonal Antibody (24GB4720)
          Product Details
          MA5-57344
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:5,000
          -
          Immunocytochemistry (ICC/IF)
          1:100-1:1,000
          -
          Flow Cytometry (Flow)
          1:2,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          24GB4720
          Immunogen
          A synthesized peptide derived from human ATE1 (aa 39-52)
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1.2 mg/mL
          Amount
          24 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          RRID
          AB_3680369
          Target Information
          Arginyl-tRNA-protein transferase (ATE1), also designated arginyltransferase 1, belongs to the R-transferase family of proteins. In order for a protein to be degraded via the ubiquitin pathway, arginylation of the protein is required. ATE1 plays an important role in this process, as it is important for the posttranslational conjugation of arginine to the N-terminal aspartate-, glutamate- and possibly cystine-contaning substrates. ATE1 is a 518 amino acid protein. Alternative splicing results in two distinct isoforms. ATE1, which is found as a monomer, can localize to the cytoplasm and/or the nucleus.
          Bioinformatics
          Protein Aliases: alternatively spliced; Arginine-tRNA--protein transferase 1; arginine-tRNA-protein transferase 1; Arginyl-tRNA--protein transferase 1; arginyl-tRNA-protein transferase; Arginyltransferase 1; R-transferase 1; unnamed protein product
          View more View less
          Gene Aliases: AI225793; ATE1; AW547406
          View more View less
          UniProt ID: (Human) O95260, (Mouse) Q9Z2A5
          View more View less
          Entrez Gene ID: (Human) 11101, (Mouse) 11907, (Rat) 293526
          View more View less
          Function(s)
          arginyltransferase activity protein binding transferase activity transferase activity, transferring acyl groups translational protein
          Process(es)
          ubiquitin-dependent protein catabolic process response to oxidative stress proteasomal protein catabolic process protein arginylation protein K27-linked ubiquitination protein K63-linked ubiquitination protein targeting to vacuole involved in autophagy ubiquitin-dependent protein catabolic process via the N-end rule pathway
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline