    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
          • Primary Antibodies ›
          • Cyclin D2 Antibodies
          Invitrogen
          Cyclin D2 Recombinant Rabbit Monoclonal Antibody (5B4M7)
          View all (33) Cyclin D2 antibodies
          Tech Support
          Cite Cyclin D2 Recombinant Rabbit Monoclonal Antibody (5B4M7)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Cyclin D2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Cyclin D2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Cyclin D2 Antibody (MA5-58016) in WB
          Western blot analysis of Cyclin D2 in U-2 OS cell lysates. Samples were incubated with Cyclin D2 Recombinant Monoclonal Antibody (Product # MA5-58016) using a dilution of 1:7, 000. Followed by a HRP-conjugated Goat anti-Rabbit IgG (H+L) Secondary Antibody at 1:10,000 dilution. Blocking buffer: 3% nonfat dry milk in TBST; Exposure: 90 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Cyclin D2 Antibody in Western Blot (WB)
          Cyclin D2 Recombinant Rabbit Monoclonal Antibody (5B4M7)
          Product Details
          MA5-58016
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:3,500-1:7,000
          -
          ELISA (ELISA)
          1 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          5B4M7
          Immunogen
          A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Cyclin D2 (NP_001750.1).
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.63 mg/mL
          Amount
          63 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.05% BSA, 50% glycerol
          Contains
          0.09% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          RRID
          AB_3681038
          Product Specific Information
          Immunogen sequence: LLGAVCMFL ASKLKETSPL TAEKLCIYTD NSIKPQELLE WELVVLGKLK WNLAAVTPHD FIEHILRKLP QQREKLSLIR KHAQTFIALC ATDFKFAMYP PS
          Target Information
          Cyclin D2 is a G1 cyclin required for G1 phase progression and is a strong candidate for a proto-oncogene. Cyclin D2 can phosphorylate pRB when associated with cdk 4 and / or cdk 6.
          Bioinformatics
          Protein Aliases: G1/S-specific cyclin-D2; KIAK0002; MGC102758; unnamed protein product
          View more View less
          Gene Aliases: CCND2; KIAK0002; MPPH3
          View more View less
          UniProt ID: (Human) P30279
          View more View less
          Entrez Gene ID: (Human) 894
          View more View less
          Function(s)
          protein binding cyclin-dependent protein serine/threonine kinase regulator activity protein kinase binding protein serine/threonine kinase activator activity cyclin-dependent protein serine/threonine kinase activator activity
          Process(es)
          G1/S transition of mitotic cell cycle long-term memory positive regulation of cell proliferation adult locomotory behavior negative regulation of apoptotic process cell division regulation of cell cycle cellular response to X-ray positive regulation of G1/S transition of mitotic cell cycle positive regulation of mitotic cell cycle phase transition regulation of G1/S transition of mitotic cell cycle
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          India flag icon
          India
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline