    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
          • Primary Antibodies ›
          • PLXNB3 Antibodies
          Invitrogen
          PLXNB3 Monoclonal Antibody (905805)
          View all (7) PLXNB3 antibodies
          Tech Support
          Cite PLXNB3 Monoclonal Antibody (905805)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          PLXNB3 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          PLXNB3 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          PLXNB3 Antibody (MA5-61503) in Flow
          Flow Cytometry analysis of PLXNB3 in A172 Human Cell Line using PLXNB3 monoclonal antibody (Product # MA5-61503). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PLXNB3 Antibody in Flow Cytometry (Flow)
          PLXNB3 Monoclonal Antibody (905805)
          Product Details
          MA5-61503
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          905805
          Immunogen
          Chinese hamster ovary cell line CHO-derived recombinant human Plexin B3, His45-Gln1255
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS with trehalose
          Contains
          no preservative
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3702115
          Product Specific Information
          Reconstitute at 0.5 mg/mL in sterile PBS.
          Target Information
          The protein encoded by this gene is a member of the plexin family. It functions as a receptor for semaphorin 5A, and plays a role in axon guidance, invasive growth and cell migration. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
          Bioinformatics
          Protein Aliases: FLJ76953; plexin 6; Plexin-B3; putative cell adhesion molecule; unnamed protein product
          View more View less
          Gene Aliases: KIAA1206; PLEXB3; PLEXR; PLXN6; PLXNB3
          View more View less
          UniProt ID: (Human) Q9ULL4
          View more View less
          Entrez Gene ID: (Human) 5365
          View more View less
          Function(s)
          protein binding semaphorin receptor activity protein domain specific binding Rho GDP-dissociation inhibitor binding protein binding involved in cell-cell adhesion
          Process(es)
          positive regulation of endothelial cell proliferation homophilic cell adhesion via plasma membrane adhesion molecules negative regulation of cell adhesion nervous system development synapse assembly regulation of cell shape negative regulation of lamellipodium assembly positive regulation of neuron projection development negative regulation of cell migration negative regulation of GTPase activity positive regulation of axonogenesis positive chemotaxis cell chemotaxis semaphorin-plexin signaling pathway
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          India flag icon
          India
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline