    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD72 Antibodies
          Invitrogen
          CD72 Monoclonal Antibody (3F3), Alexa Fluor™ 488
          View all (28) CD72 antibodies
          Tech Support
          Cite CD72 Monoclonal Antibody (3F3), Alexa Fluor™ 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          CD72 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD72 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          CD72 Antibody (MA562340) in Flow
          Flow cytometry (surface staining) of CD72 in human peripheral whole blood. Samples were stained with CD72 monoclonal antibody (Product # MA562340) using a dilution of 4 µL reagent/100 µL of peripheral whole blood. Separation of human CD72 positive lymphocytes (red-filled) from CD72 negative lymphocytes (black-dashed). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD72 Antibody in Flow Cytometry (Flow)
          CD72 Antibody in Flow Cytometry (Flow)
          CD72 Monoclonal Antibody (3F3), Alexa Fluor™ 488
          Product Details
          MA562340
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          4 µL/100 µL whole blood or 10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3F3
          Immunogen
          Normal human lymphocytes from a lymph node.
          3D Epitope / Immunogen
          Conjugate
          Alexa Fluor™ 488 Alexa Fluor™ 488 Alexa Fluor™ 488
          • Unconjugated
          • PE 
          • Request custom conjugation
          Excitation/Emission Max
          499/520 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Purification
          Size-exclusion chromatography
          Storage buffer
          PBS, pH 7.4, with 0.2% BSA
          Contains
          15mM sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3703526
          Target Information
          CD72 is a transmembrane glycoprotein expressed as a homodimer especially in B cells, but also in other antigen presenting cells such as dendritic cells and macrophages. Through one of its immunoreceptor tyrosine-based inhibitory motives (ITIMs), CD72 interacts with tyrosine phosphatase SHP-1, thereby suppressing B cell responsiveness. Binding of CD72 with its ligand CD100 (Sema4D) prevents BCR association and phosphorylation of CD72 and results in dissociation of SHP-1 from CD72, thus enables B cell activation.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: B-cell differentiation antigen CD72; CD72; CD72 antigen; Lyb-2; one of the receptors for CD100; RP11-331F9.9
          View more View less
          Gene Aliases: CD72; CD72b; LYB2
          View more View less
          UniProt ID: (Human) P21854
          View more View less
          Entrez Gene ID: (Human) 971
          View more View less
          Function(s)
          transmembrane signaling receptor activity receptor binding protein binding carbohydrate binding
          Process(es)
          cell adhesion negative regulation of B cell receptor signaling pathway
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-5btrv:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline