    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO and CRO Solutions
          • Primary Antibodies ›
          • WBP1L Antibodies
          Invitrogen
          WBP1L Monoclonal Antibody (OPAL1-01), Alexa Fluor™ 647
          View all (5) WBP1L antibodies
          Tech Support
          Cite WBP1L Monoclonal Antibody (OPAL1-01), Alexa Fluor™ 647
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          WBP1L Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          WBP1L Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          WBP1L Antibody (MA562390) in Flow
          Flow cytometry (intracellular staining) of WBP1L in HEK-293 cells. Samples were stained with WBP1L monoclonal antibody (Product # MA562390) using a dilution of 0.6 µg/mL. Separation of stained HEK-293 cells (red-filled) from HEK-293 cells stained using mouse IgG2a isotype control (MOPC-173) Alexa Fluor® 647 antibody (0.6 µg/mL, black-dashed). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          WBP1L Antibody in Flow Cytometry (Flow)
          WBP1L Monoclonal Antibody (OPAL1-01), Alexa Fluor™ 647
          Product Details
          MA562390
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Flow Cytometry (Flow)
          0.5-4 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OPAL1-01
          Immunogen
          Recombinant fragment of human OPAL1 (amino acids 152-342).
          3D Epitope / Immunogen
          Conjugate
          Alexa Fluor™ 647 Alexa Fluor™ 647 Alexa Fluor™ 647
          • Unconjugated
          • PE 
          • Request custom conjugation
          Excitation/Emission Max
          650/671 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Amount
          100 µg
          Purification
          Size-exclusion chromatography
          Storage buffer
          PBS, pH 7.4, with 0.2% BSA
          Contains
          15mM sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3703576
          Target Information
          WBP1L (OPAL1, outcome predictor in acute leukemia 1) is an almost uncharacterized transmembrane adaptor protein expressed mainly by megakaryocytes and platelets. Its expression is enhanced on TEL/AML leukemic cells. The function of OPAL1 is unknown, although the presence of a cytochrome c-like heme-binding site and a transmembrane domain suggested OPAL1 may be involved in the mitochondrial electron transport chain. Its originally reported prognostic impact appears to be treatment dependent.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: Outcome predictor in acute leukemia 1; unnamed protein product; WW domain binding protein 1-like
          View more View less
          Gene Aliases: C10orf26; OPA1L; OPAL1; WBP1L
          View more View less
          UniProt ID: (Human) Q9NX94
          View more View less
          Entrez Gene ID: (Human) 54838
          View more View less
          Function(s)
          protein binding ubiquitin protein ligase binding
          Process(es)
          hemopoiesis positive regulation of protein ubiquitination CXCL12-activated CXCR4 signaling pathway
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline