RALGAPA1 is a catalytic subunit of the heterodimeric RalGAP1 complex which acts as a GTPase activator for the Ras-like small GTPases RALA and RALB.
Protein Aliases: GAP-related interacting partner to E12; GAP-related interacting protein to E12; GAP-related-interacting partner to E12; GRIPE; GTPase activating RANGAP domain-like 1; GTPase activating Rap/RanGAP domain-like 1; GTPase-activating Rap/Ran-GAP domain-like 1; GTPase-activating RapGAP domain-like 1; p240; Ral GTPase activating protein catalytic alpha subunit 1; Ral GTPase activating protein, alpha subunit 1 (catalytic); ral GTPase-activating protein alpha subunit 1; Ral GTPase-activating protein subunit alpha-1; Tuberin-like protein 1; tulip 1 protein; unnamed protein product
Gene Aliases: 2310003F20Rik; 4930400K19Rik; AI563624; GARNL1; GRIPE; KIAA0884; mKIAA0884; NEDHRIT; p240; RALGAPA1; RalGAPalpha1; TULIP1
UniProt ID: (Human) Q6GYQ0, (Mouse) Q6GYP7, (Rat) O55007
Entrez Gene ID: (Human) 253959, (Mouse) 56784, (Rat) 56785