    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TEM7 Antibodies
          Invitrogen
          TEM7 Recombinant Mouse Monoclonal Antibody (K9N025_3D5)
          View all (16) TEM7 antibodies
          Tech Support
          Cite TEM7 Recombinant Mouse Monoclonal Antibody (K9N025_3D5)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          TEM7 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TEM7 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          TEM7 Antibody (MA564455) in WB
          Western blot analysis of TEM7 in rat liver tissue lysate (15 µg). Samples were incubated with TEM7 recombinant monoclonal (Product # MA564455) using a dilution of 1 µg/mL followed by peroxidase conjugated rabbit anti-mouse IgG (Light chain specific). Sample was run on 6-18% SDS-PAGE under reducing conditions and blotted onto nitrocellulose membrane. PLXDC1 band was visualized using ECL Western Blotting Substrate. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TEM7 Antibody in Western Blot (WB)
          TEM7 Recombinant Mouse Monoclonal Antibody (K9N025_3D5)
          Product Details
          MA564455
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Mouse / IgG2a
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          K9N025_3D5
          Immunogen
          Recombinant human PLXDC1 (aa 341-420).
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS with 0.5% BSA, 50% glycerol
          Contains
          0.09% sodium azide
          Storage conditions
          Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3705542
          Product Specific Information
          Immunogen sequence: EWMDYGCAQE AEGRMCEDFQ DEDHDSASPD TSFSPYDGDL TTTSSSLFID SLTTEDDTKL NPYAGGDGLQ NNLSPKTKGT
          Highest antigen sequence identity to the following orthologs: Mouse - 80%, Pig - 78%, Cynomolgus monkey - 96%.
          Target Information
          Recently, using SAGE (Serial Analysis of Gene Expression) technology, St. Croix et al, have identified 46 genes, whose expression is specifically elevated in tumor-associated endothelium. Nine of these genes were prominently expressed only in tumor endothelial cells (EC), but were absent or barely detectable in normal ECs, and named as Tumor Endothelial Markers (TEMs, TEM 1-9). TEM7 (Tumor endothelial marker 7) transcripts are specifically expressed in the endothelium of colorectal cancer, primary cancers of lung, pancreas, breast, and brain. TEM7 is expressed specifically in endothelium of these cancers, whether primary or metastasis. The other six members of this family (TEM1, 3, 4, 5, 8, and 9) also show similar expression pattern in lung and brain tumors, and liver metastasis. Since most of the genes expressed differentially in tumor endothelium are also expressed during angiogenesis, these newly discovered genes might provide important resources for basic and clinical studies of human angiogenesis.
          Bioinformatics
          Protein Aliases: 2410003I07Rik; ADP-ribosylation factor-like 12; alternatively spliced; DKFZp686F0937; FLJ36270; FLJ45632; Plexin domain-containing protein 1; TEM7 precursor; Tumor endothelial marker 3; Tumor endothelial marker 7; tumor endothelial marker7; unnamed protein product
          View more View less
          Gene Aliases: Arl12; PLXDC1; TEM3; TEM7
          View more View less
          UniProt ID: (Human) Q8IUK5
          View more View less
          Entrez Gene ID: (Human) 57125, (Rat) 303505
          View more View less
          Function(s)
          protein binding molecular_function receptor activity
          Process(es)
          angiogenesis spinal cord development
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
          git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.49.0-Offline