    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Flotillin 2 Antibodies
          Invitrogen
          Flotillin 2 Recombinant Rabbit Monoclonal Antibody (K01_3M89)
          View all (18) Flotillin 2 antibodies
          Tech Support
          Cite Flotillin 2 Recombinant Rabbit Monoclonal Antibody (K01_3M89)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Flotillin 2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Flotillin 2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Flotillin 2 Antibody (MA566674) in WB
          Western Blot analysis of Flotillin 2 in Lane 1: Hela lysates; Lane 2: 293 lysates using Flotillin 2 Recombinant Monoclonal Antibody (Product # MA566674). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Flotillin 2 Antibody in Western Blot (WB)
          Product Details
          MA566674
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          K01_3M89
          Immunogen
          A synthesized peptide derived from human Flotillin 2.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4, with 50% glycerol, 0.05% BSA, 150mM NaCl
          Contains
          0.02% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. This gene encodes a caveolae-associated, integral membrane protein, which is thought to function in neuronal signaling.
          Bioinformatics
          Protein Aliases: Epidermal surface antigen; ESA; FLOT2; Flotillin 2 (epidermal surface antigen 1); Flotillin-2; Membrane component chromosome 17 surface marker 1; Membrane component chromosome 17 surface marker 1 homolog; membrane component, chromosome 17, surface marker 1 (35kD protein identified by monoclonal antibody ECS-1); OTTHUMP00000163513; REG-1; Reggie-1; reggie1-1
          View more View less
          Gene Aliases: AI573412; ECS-1; ECS1; ESA; ESA1; FLOT2; M17S1; Reg1; reggie-2
          View more View less
          UniProt ID: (Human) Q14254, (Mouse) Q60634, (Rat) Q9Z2S9
          View more View less
          Entrez Gene ID: (Human) 2319, (Mouse) 14252, (Rat) 83764
          View more View less
          Function(s)
          protease binding protein binding ionotropic glutamate receptor binding protein heterodimerization activity
          Process(es)
          membrane raft assembly cell adhesion negative regulation of gene expression protein localization to plasma membrane raft protein stabilization establishment of protein localization to plasma membrane negative regulation of amyloid precursor protein catabolic process positive regulation of establishment of T cell polarity epidermis development positive regulation of cell-cell adhesion positive regulation of I-kappaB kinase/NF-kappaB signaling regulation of myoblast differentiation protein localization to plasma membrane anterograde dendritic transport regulation of postsynaptic membrane neurotransmitter receptor levels
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline