    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
          • Order Status
          • 快速訂購
          • 登入
            登入
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • ADH1A Antibodies
          Invitrogen
          Alcohol Dehydrogenase 1A Recombinant Rabbit Monoclonal Antibody (K01_3Y30)
          View all (16) ADH1A antibodies
          Tech Support
          Cite Alcohol Dehydrogenase 1A Recombinant Rabbit Monoclonal Antibody (K01_3Y30)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Alcohol Dehydrogenase 1A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Alcohol Dehydrogenase 1A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Alcohol Dehydrogenase 1A Antibody (MA567156) in WB
          Western Blot analysis of Alcohol Dehydrogenase 1A in HepG2 lysates using Alcohol Dehydrogenase 1A Recombinant Monoclonal Antibody (Product # MA567156). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Alcohol Dehydrogenase 1A Antibody in Western Blot (WB)
          Product Details
          MA567156
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          K01_3Y30
          Immunogen
          A synthesized peptide derived from human Alcohol Dehydrogenase.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4, with 50% glycerol, 150mM NaCl, 0.05% BSA
          Contains
          0.02% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          This gene encodes a member of the alcohol dehydrogenase family. The encoded protein is the alpha subunit of class I alcohol dehydrogenase, which consists of several homo- and heterodimers of alpha, beta and gamma subunits. Alcohol dehydrogenases catalyze the oxidation of alcohols to aldehydes. This gene is active in the liver in early fetal life but only weakly active in adult liver. This gene is found in a cluster with six additional alcohol dehydrogenase genes, including those encoding the beta and gamma subunits, on the long arm of chromosome 4. Mutations in this gene may contribute to variation in certain personality traits and substance dependence.
          Bioinformatics
          Protein Aliases: ADH, alpha subunit; ADH-A2; Alcohol dehydrogenase 1; alcohol dehydrogenase 1 (class I), alpha polypeptide; alcohol dehydrogenase 1 temporal, liver; alcohol dehydrogenase 1, complex; alcohol dehydrogenase 1, electrophoretic; Alcohol dehydrogenase 1A; alcohol dehydrogenase 3, electrophoretic; Alcohol dehydrogenase A subunit; alcohol dehydrogenase alpha subunit (EC 1.1.1.1); Alcohol dehydrogenase subunit alpha; aldehyde reductase; class I alcohol dehydrogenase; unnamed protein product
          View more View less
          Gene Aliases: Adh; Adh-1; Adh-1-t; Adh-1e; Adh-1t; Adh-3e; ADH-AA; ADH1; Adh1-e; Adh1-t; ADH1A; Adh1c; Adh1tl; Adh3-e; AI194826
          View more View less
          UniProt ID: (Human) P07327, (Mouse) P00329, (Rat) P06757
          View more View less
          Entrez Gene ID: (Human) 124, (Mouse) 11522, (Rat) 24172
          View more View less
          Function(s)
          alcohol dehydrogenase (NAD) activity retinol dehydrogenase activity protein binding zinc ion binding oxidoreductase activity metal ion binding butanol dehydrogenase (NAD+) activity drug binding ethanol binding protein homodimerization activity NAD binding dehydrogenase oxidoreductase metabolite interconversion enzyme
          Process(es)
          alcohol metabolic process retinol metabolic process retinoic acid metabolic process retinoid metabolic process ethanol catabolic process ethanol oxidation response to retinoic acid response to testosterone acetaldehyde biosynthetic process behavioral response to ethanol response to steroid hormone oxidation-reduction process
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline