          • Primary Antibodies ›
          • SLC25A5 Antibodies
          Invitrogen
          SLC25A5 Recombinant Rabbit Monoclonal Antibody (9S4V9)
          View all (13) SLC25A5 antibodies
          Tech Support
          Cite SLC25A5 Recombinant Rabbit Monoclonal Antibody (9S4V9)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          SLC25A5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SLC25A5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          SLC25A5 Antibody (MA569092) in ICC/IF
          Immunocytochemical analysis of SLC25A5 in MCF7 cells. Samples were incubated with SLC25A5 recombinant monoclonal antibody (Product # MA569092) using a dilution of 1:100 (40x lens). Secondary antibody: Cy3-conjugated Goat anti-Rabbit IgG (H+L) using a dilution of 1:500. DAPI for nuclear staining (Blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SLC25A5 Antibody in Immunocytochemistry (ICC/IF)
          SLC25A5 Antibody in Immunocytochemistry (ICC/IF)
          SLC25A5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SLC25A5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SLC25A5 Antibody in Western Blot (WB)
          Product Details
          MA569092
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:5,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:100-1:500
          -
          Immunocytochemistry (ICC/IF)
          1:50-1:200
          -
          ELISA (ELISA)
          1 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          9S4V9
          Immunogen
          Recombinant protein (or fragment).
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1.09 mg/mL
          Amount
          109 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3., with 50% glycerol, 0.05% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          Adenine nucleotide translocator (ANT) and the voltage-dependent anion-selective channel proteins 1 and 2 (VDAC1 and VDAC2) are components of the permeability transition pore complex (PTPC) of the mitochondrial inner and outer membranes, respectively. Formation of PTPCs, the subsequent dissipation of mitochondrial inner membrane potential and release of cytochrome c through the outer mitochondrial membrane are critical events in the early stages of apoptosis. Bax, a proapoptotic protein, has been shown to act upon ANT to induce the dissipation of mitochondrial inner membrane potential. ANT1 has a role in the maintenance of mitochondrial DNA by catalyzing the exchange of ADP and ATP across the mitochondrial inner membrane.
          Bioinformatics
          Protein Aliases: Adenine nucleotid translocator 2 fibroblast isoform (ATP-ADP carrier protein); Adenine nucleotid translocator 2, fibroblast isoform (ATP-ADP carrier protein); Adenine nucleotide translocator 2; adenine nucleotide translocator 2 (fibroblast); adenine nucleotide translocator 2 fibroblast isoform (ATP-ADP carrier protein); adenine nucleotide translocator 2, fibroblast isoform (ATP-ADP carrier protein); ADP,ATP carrier protein 2; ADP,ATP carrier protein, fibroblast isoform; ADP.ATP translocase; ADP/ATP carrier protein; ADP/ATP translocase 2; ANT 2; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 5; Solute carrier family 25 member 5; unnamed protein product
          View more View less
          Gene Aliases: 2F1; AAC2; ANT2; SLC25A5; T2; T3
          View more View less
          UniProt ID: (Human) P05141, (Rat) Q09073, (Mouse) P51881
          View more View less
          Entrez Gene ID: (Human) 292, (Rat) 25176, (Mouse) 11740
          View more View less
          Function(s)
          adenine nucleotide transmembrane transporter activity RNA binding ATP:ADP antiporter activity protein binding hydrogen ion transmembrane transporter activity adenine transmembrane transporter activity antiporter activity oxidative phosphorylation uncoupler activity ubiquitin protein ligase binding transporter activity poly(A) RNA binding transfer/carrier protein
          Process(es)
          chromosome segregation positive regulation of cell proliferation adenine transport B cell differentiation erythrocyte differentiation regulation of mitochondrial membrane permeability adenine nucleotide transport transmembrane transport mitochondrial ADP transmembrane transport negative regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway positive regulation of mitophagy hydrogen ion transmembrane transport mitochondrial ATP transmembrane transport cellular response to leukemia inhibitory factor adaptive thermogenesis
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline