    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
          • Primary Antibodies ›
          • ATP Citrate Lyase Antibodies
          Invitrogen
          Phospho-ATP Citrate Lyase (Ser455) Recombinant Rabbit Monoclonal Antibody (6S0P10)
          View all (40) ATP Citrate Lyase antibodies
          Tech Support
          Cite Phospho-ATP Citrate Lyase (Ser455) Recombinant Rabbit Monoclonal Antibody (6S0P10)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (7)
          Phospho-ATP Citrate Lyase (Ser455) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Phospho-ATP Citrate Lyase (Ser455) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 7
          Phospho-ATP Citrate Lyase (Ser455) Antibody (MA569900) in IHC (P)
          Immunohistochemistry analysis of Phospho-ATP Citrate Lyase (Ser455) on paraffin-embedded Human placenta tissue. Samples were incubated with Phospho-ATP Citrate Lyase (Ser455) recombinant monoclonal antibody (Product # MA569900) using a dilution of 1:200 (40x lens). High pressure antigen retrieval was performed with 0. 01 M citrate buffer (pH 6. 0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Phospho-ATP Citrate Lyase (Ser455) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-ATP Citrate Lyase (Ser455) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-ATP Citrate Lyase (Ser455) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-ATP Citrate Lyase (Ser455) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-ATP Citrate Lyase (Ser455) Antibody in Western Blot (WB)
          Phospho-ATP Citrate Lyase (Ser455) Antibody in Western Blot (WB)
          Phospho-ATP Citrate Lyase (Ser455) Antibody in Western Blot (WB)
          Product Details
          MA569900
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:3,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:200
          -
          ELISA (ELISA)
          1 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          6S0P10
          Immunogen
          Synthetic peptide.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1.58 mg/mL
          Amount
          158 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3., with 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          ATP citrate lyase is the primary enzyme responsible for the synthesis of cytosolic acetyl-CoA in many tissues. The enzyme is a tetramer (relative molecular weight approximately 440,000) of apparently identical subunits. It catalyzes the formation of acetyl-CoA and oxaloacetate from citrate and CoA with a concomitant hydrolysis of ATP to ADP and phosphate. The product, acetyl-CoA, serves several important biosynthetic pathways, including lipogenesis and cholesterogenesis. In nervous tissue, ATP citrate-lyase may be involved in the biosynthesis of acetylcholine. Two transcript variants encoding distinct isoforms have been identified for this gene.
          Bioinformatics
          Protein Aliases: ACL; ATP-citrate (pro-S-)-lyase; ATP-citrate synthase; Citrate cleavage enzyme; unnamed protein product
          View more View less
          Gene Aliases: A730098H14Rik; ACL; ACLY; ATPCL; AW538652; CLATP
          View more View less
          UniProt ID: (Human) P53396, (Mouse) Q91V92, (Rat) P16638
          View more View less
          Entrez Gene ID: (Human) 47, (Mouse) 104112, (Rat) 24159
          View more View less
          Function(s)
          nucleotide binding catalytic activity ATP citrate synthase activity protein binding ATP binding transferase activity metal ion binding transferase activity, transferring acyl groups, acyl groups converted into alkyl on transfer succinate-CoA ligase (ADP-forming) activity cofactor binding lyase activity transferase
          Process(es)
          acetyl-CoA biosynthetic process citrate metabolic process oxaloacetate metabolic process lipid metabolic process fatty acid biosynthetic process cholesterol biosynthetic process lipid biosynthetic process coenzyme A metabolic process negative regulation of ferroptosis acetyl-CoA metabolic process metabolic process cellular carbohydrate metabolic process
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          India flag icon
          India
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline