    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • HIP2 Antibodies
          Invitrogen
          HIP2 Recombinant Rabbit Monoclonal Antibody (PSH11-24)
          View all (13) HIP2 antibodies
          Tech Support
          Cite HIP2 Recombinant Rabbit Monoclonal Antibody (PSH11-24)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (12)
          HIP2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          HIP2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 12
          HIP2 Antibody (MA569964) in ICC/IF
          Immunocytochemistry analysis of HIP2 in NIH/3T3 cells. Samples were incubated with HIP2 monoclonal antibody (Product # MA569964) using a dilution of 1:100 in 1% BSA in PBST overnight at 4°C followed by Goat Anti-Rabbit IgG H&L (iFluor™ 488) at a dilution of 1:1,000. Cells were fixed in 4% paraformaldehyde for 15 minutes at room temperature, permeabilized with 0.1% Triton X-100 in PBS for 15 minutes at room temperature, then blocked with 1% BSA in 10% negative goat serum for 1 hour at room temperature. PBS instead of the primary antibody wa... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          HIP2 Antibody in Immunocytochemistry (ICC/IF)
          HIP2 Antibody in Immunocytochemistry (ICC/IF)
          HIP2 Antibody in Immunocytochemistry (ICC/IF)
          HIP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HIP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HIP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HIP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HIP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HIP2 Antibody in Western Blot (WB)
          HIP2 Antibody in Flow Cytometry (Flow)
          HIP2 Antibody in Flow Cytometry (Flow)
          HIP2 Antibody in Flow Cytometry (Flow)
          HIP2 Recombinant Rabbit Monoclonal Antibody (PSH11-24)
          Product Details
          MA569964
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:1,000
          -
          Immunocytochemistry (ICC/IF)
          1:100
          -
          Flow Cytometry (Flow)
          1:1,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Non-human primate, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          PSH11-24
          Immunogen
          Recombinant protein within human HIP2 aa 1-200.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.4, with 0.1% BSA, 40% glycerol
          Contains
          0.05% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Positive control: HeLa cell lysate, 293T cell lysate, HCT 116 cell lysate, Daudi cell lysate, Jurkat cell lysate, K-562 cell lysate, U-2 OS cell lysate, COS-1 cell lysate, C2C12 cell lysate, NIH/3T3 cell lysate, PC-12 cell lysate, Mouse brain tissue lysate, Rat brain tissue lysate, human brain tissue, human liver tissue, mouse heart tissue, mouse liver tissue, rat liver tissue.
          Subcellular location: Cytoplasm.
          Target Information
          Accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In vitro, in the presence or in the absence of BRCA1-BARD1 E3 ubiquitin-protein ligase complex, catalyzes the synthesis of 'Lys-48'-linked polyubiquitin chains. Does not transfer ubiquitin directly to but elongates monoubiquitinated substrate protein. Mediates the selective degradation of short-lived and abnormal proteins, such as the endoplasmic reticulum-associated degradation (ERAD) of misfolded lumenal proteins. Ubiquitinates huntingtin. May mediate foam cell formation by the suppression of apoptosis of lipid-bearing macrophages through ubiquitination and subsequence degradation of p53/TP53. Proposed to be involved in ubiquitination and proteolytic processing of NF-kappa-B; in vitro supports ubiquitination of NFKB1. In case of infection by cytomegaloviruses may be involved in the US11-dependent degradation of MHC class I heavy chains following their export from the ER to the cytosol. In case of viral infections may be involved in the HPV E7 protein-dependent degradation of RB1.
          Bioinformatics
          Protein Aliases: DKFZp564C1216; DKFZp686J24237; E2 ubiquitin-conjugating enzyme K; E2(25K); HIP-2; huntingtin interacting protein 2; Huntingtin-interacting protein 2; Ubiquitin carrier protein; ubiquitin conjugating enzyme E2K; Ubiquitin-conjugating enzyme E2 K; Ubiquitin-conjugating enzyme E2(25K); Ubiquitin-conjugating enzyme E2-25 kDa; ubiquitin-conjugating enzyme E2-25K; ubiquitin-conjugating enzyme E2K (UBC1 homolog, yeast); Ubiquitin-protein ligase; unnamed protein product
          View more View less
          Gene Aliases: AW492011; D5Ertd601e; E2-25K; HIP2; HYPG; LIG; UBC1; UBE2K
          View more View less
          UniProt ID: (Human) P61086, (Mouse) P61087
          View more View less
          Entrez Gene ID: (Human) 3093, (Mouse) 53323, (Rat) 289623
          View more View less
          Function(s)
          nucleotide binding ubiquitin-protein transferase activity protein binding ATP binding transferase activity ubiquitin protein ligase binding ubiquitin-ubiquitin ligase activity ubiquitin conjugating enzyme activity ligase activity ubiquitin protein ligase activity ubiquitin-like protein transferase activity ubiquitin-protein ligase
          Process(es)
          protein polyubiquitination ubiquitin-dependent protein catabolic process positive regulation of peptidyl-threonine phosphorylation free ubiquitin chain polymerization protein ubiquitination regulation of proteasomal ubiquitin-dependent protein catabolic process cellular response to interferon-beta type I interferon signaling pathway positive regulation of type I interferon-mediated signaling pathway protein K48-linked ubiquitination positive regulation of tumor necrosis factor-mediated signaling pathway proteasome-mediated ubiquitin-dependent protein catabolic process intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline