    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • IL4R Antibodies
          Invitrogen
          IL4R Recombinant Rabbit Monoclonal Antibody (PSH10-86)
          View all (75) IL4R antibodies
          Tech Support
          Cite IL4R Recombinant Rabbit Monoclonal Antibody (PSH10-86)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          IL4R Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          IL4R Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          IL4R Antibody (MA570137) in ICC/IF
          Immunocytochemistry analysis of IL4R in PC-12 cells. Samples were incubated with IL4R monoclonal antibody (Product # MA570137) using a dilution of 1:100 in 1% BSA in PBST overnight at 4°C followed by Goat Anti-Rabbit IgG H&L (iFluor™ 488) at a dilution of 1:1,000. Cells were fixed in 4% paraformaldehyde for 15 minutes at room temperature, permeabilized with 0.1% Triton X-100 in PBS for 15 minutes at room temperature, then blocked with 1% BSA in 10% negative goat serum for 1 hour at room temperature. PBS instead of the primary antibody was ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          IL4R Antibody in Immunocytochemistry (ICC/IF)
          IL4R Antibody in Western Blot (WB)
          IL4R Antibody in Immunoprecipitation (IP)
          IL4R Recombinant Rabbit Monoclonal Antibody (PSH10-86)
          Product Details
          MA570137
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000
          -
          Immunocytochemistry (ICC/IF)
          1:100
          -
          Immunoprecipitation (IP)
          1-2 µg
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          PSH10-86
          Immunogen
          Recombinant protein within human IL-4R aa 1-232.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.4, with 0.1% BSA, 40% glycerol
          Contains
          0.05% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Positive control: THP-1 cell lysate, Daudi cell lysate, 293T cell lysate, K-562 cell lysate, Jurkat cell lysate, MCF7 cell lysate, Ramos cell lysate, RAW264.7 cell lysate, PC-12 cell lysate, Mouse spleen tissue lysate, Mouse pancreas tissue lysate.
          Subcellular location: Cell membrane; Secreted.
          Target Information
          IL4 Receptor alpha (IL4RA) is the alpha chain of the interleukin 4 receptor which binds to both interleukin 4 and interleukin 13 to regulate IgE production, chemokine and mucus production at sites of allergic inflammmation. The IL4 response is involved in promoting Th2 differentiation. The secreted extracellular domain of IL4R alpha, called sIL4R alpha, can inhibit IL4-mediated cell proliferation and IL5 upregulation by T-cells. It has no signaling abilities. Allelic variations in the IL4RA gene have been associated with atopy, a condition that can manifest itself as allergic rhinitis, sinusitus, asthma, or eczema.
          Bioinformatics
          Protein Aliases: CD124; il-4 receptor; IL-4 receptor alpha chain; IL-4 receptor subunit alpha; IL-4R subunit alpha; IL-4R-alpha; IL-4RA; IL4Ra; interleukin 13 receptor; interleukin 4 receptor, alpha; interleukin-4 receptor alpha chain; Interleukin-4 receptor subunit alpha; unnamed protein product
          View more View less
          Gene Aliases: 582J2.1; CD124; IL-4RA; IL4R; IL4RA
          View more View less
          UniProt ID: (Human) P24394, (Rat) Q63257, (Mouse) P16382
          View more View less
          Entrez Gene ID: (Human) 3566, (Rat) 25084, (Mouse) 16190
          View more View less
          Function(s)
          cytokine receptor activity interleukin-4 receptor activity protein binding protein heterodimerization activity transmembrane signal receptor
          Process(es)
          immune system process production of molecular mediator involved in inflammatory response positive regulation of immunoglobulin production immune response signal transduction immunoglobulin mediated immune response cytokine-mediated signaling pathway positive regulation of chemokine production interleukin-4-mediated signaling pathway defense response to protozoan positive regulation of macrophage activation positive regulation of mast cell degranulation T-helper 1 cell differentiation T-helper 2 cell differentiation negative regulation of T-helper 1 cell differentiation positive regulation of T-helper 2 cell differentiation positive regulation of cold-induced thermogenesis positive regulation of myoblast fusion inflammatory response ovulation regulation of cell proliferation response to estrogen positive regulation of chemokine secretion
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline