    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • TRIM Antibodies
          Invitrogen
          TRIM Monoclonal Antibody (TRIM-04), Alexa Fluor™ 647
          View all (8) TRIM antibodies
          Tech Support
          Cite TRIM Monoclonal Antibody (TRIM-04), Alexa Fluor™ 647
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          TRIM Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          TRIM Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          TRIM Antibody (MA574241) in Flow
          Flow cytometry analysis of TRIM in human CD3 positive TRIM positive T cells (red-filled); CD3 negative TRIM negative lymphocytes (black-dashed). Samples were incubated with monoclonal TRIM (Product # MA574241) at a dilution of 1.67 μg/mL, followed by Alexa Fluor 647. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TRIM Antibody in Flow Cytometry (Flow)
          TRIM Antibody in Flow Cytometry (Flow)
          TRIM Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          TRIM Monoclonal Antibody (TRIM-04), Alexa Fluor™ 647
          Product Details
          MA574241
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          1-5 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          TRIM-04
          Immunogen
          Recombinant intracellular domain (aa 29-186) of human TRIM.
          3D Epitope / Immunogen
          Conjugate
          Alexa Fluor™ 647 Alexa Fluor™ 647 Alexa Fluor™ 647
          • Unconjugated
          • Biotin 
          • PE 
          • Request custom conjugation
          Excitation/Emission Max
          650/671 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Amount
          100 µg
          Purification
          Size-exclusion chromatography
          Storage buffer
          PBS, pH 7.4, with 0.2% BSA
          Contains
          15mM sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          TRIM (T cell receptor-interacting molecule), also known as TRAT1 (T cell receptor associated transmembrane adaptor 1) is a 30 kDa protein expressed by T cells as a cystein-linked homodimer. It associates with TCR-CD3-zeta-chain complex and becomes phosphorylated by Src-family kinases. TRIM is potentially involved in negative regulation of TCR-mediated signaling, but its role remains unclear.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: T cell receptor interacting molecule; unnamed protein product
          View more View less
          Gene Aliases: HSPC062; pp29/30; TCRIM; TRIM
          View more View less
          UniProt ID: (Human) Q6PIZ9
          View more View less
          Entrez Gene ID: (Human) 50852
          View more View less
          Function(s)
          transmembrane receptor protein tyrosine kinase adaptor activity protein binding
          Process(es)
          negative regulation of receptor recycling cellular defense response signal transduction transmembrane receptor protein tyrosine kinase signaling pathway positive regulation of calcium-mediated signaling positive regulation of T cell receptor signaling pathway negative regulation of transport
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-74d7b4b4d9-vlj45:80/100.66.77.38:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline