    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • SCIMP Antibodies
          Invitrogen
          SCIMP Monoclonal Antibody (NVL-07), PE
          View all (4) SCIMP antibodies
          Tech Support
          Cite SCIMP Monoclonal Antibody (NVL-07), PE
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          SCIMP Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          SCIMP Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          SCIMP Antibody (MA574359) in Flow
          Flow cytometry analysis of SCIMP in human HLA-DR/DP positive SCIMP positive lymphocytes (red-filled) from HLA-DR/DP negative SCIMP negative lymphocytes (black-dashed). Samples were incubated with monoclonal SCIMP (Product # MA574359) using a dilution of 10 µL reagent/100 µL of peripheral whole blood, followed by PE. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SCIMP Antibody in Flow Cytometry (Flow)
          SCIMP Antibody in Flow Cytometry (Flow)
          SCIMP Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          MA574359
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          10 µL/10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          NVL-07
          Immunogen
          Recombinant intracellular part of human SCIMP.
          Conjugate
          PE PE PE
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          565/576 nm View spectra spectra
          Form
          Liquid
          Purification
          Size-exclusion chromatography
          Storage buffer
          PBS, pH 7.4, with 0.2% BSA
          Contains
          15mM sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          SCIMP (SLP adaptor and Csk interacting membrane protein), also known as Nvl, is a palmitoylated transmembrane adaptor protein expressed in professional antigen presenting cells, most prominently in the lymph nodes and spleen. It is associated with tetraspanin-enriched microdomains (together with MHC II). There is a close relationship between SCIMP and tyrosinkinase Lyn, which is constitutively bound to it by its SH3 domain. After MHC II-mediated stimulation in the immunological synapse SCIMP becomes phosphorylated at several tyrosine residues and provides docking sites for Grb2 and SLP65 or SLP76 adaptors transducing the signal downstream, as well as for the kinase Csk with modulatory roles.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: DTFT5783; SLP65/SLP76, Csk-interacting membrane protein; transmembrane protein C17orf87; unnamed protein product
          View more View less
          Gene Aliases: C17orf87; UNQ5783
          View more View less
          UniProt ID: (Human) Q6UWF3
          View more View less
          Entrez Gene ID: (Human) 388325
          View more View less
          Function(s)
          protein binding binding, bridging
          Process(es)
          positive regulation of cytokine production involved in immune response positive regulation of dendritic cell cytokine production positive regulation of lipopolysaccharide-mediated signaling pathway positive regulation of interleukin-12 production positive regulation of interleukin-6 production positive regulation of stress-activated MAPK cascade toll-like receptor 3 signaling pathway toll-like receptor 4 signaling pathway toll-like receptor 7 signaling pathway toll-like receptor TLR1:TLR2 signaling pathway positive regulation of ERK1 and ERK2 cascade cellular response to molecule of fungal origin
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-green-7bf6cff7cf-x9r6b:80/100.66.78.146:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0