FIGURE: 1 / 3
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
CD41 is a transmembrane glycoprotein (integrin family) composed of two chains GPIIb alpha (heavy chain; 120 kDa) and GPIIb beta (light chain; 23 kDa). CD61 non-covalently associates with CD41.
Protein Aliases: alphaIIb protein; antigen CD61; glycoprotein IIIa precursor; GPalpha IIb; Integrin alpha-2b; integrin beta 3; integrin beta chain, beta 3; integrin beta-3; integrin beta-3 subunit; integrin homolog; integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41); integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); plate glycoprotein IIIa (GPIIIa); platelet fibrinogen receptor, alpha subunit; platelet glycoprotein IIb (GPIIb); platelet glycoprotein IIb of IIb/IIIa complex; platelet membrane glycoprotein IIb; platelet membrane glycoprotein IIIa; platelet-specific antigen BAK; protein phosphatase 1, regulatory subunit 93; unnamed protein product
Gene Aliases: BDPLT16; BDPLT2; BDPLT24; CD41; CD41B; CD61; FMAIT1; FMAIT2; GP2B; GP3A; GPIIb; GPIIIa; GT; GT1; GT2; GTA; HPA3; PPP1R93
UniProt ID: (Human) P08514, (Human) P05106
Entrez Gene ID: (Human) 3674, (Human) 3690