Thermo Fisher Scientific Logo
๋กœ๊ทธ์ธ
ํšŒ์›์ด ์•„๋‹ˆ์‹ ๊ฐ€์š”? ๊ณ„์ • ์ƒ์„ฑํ•˜๊ธฐ
  • ๋ชจ๋“  ์ œํ’ˆ ์ฃผ๋ฌธ
    • ํ•ญ์ฒด
    • TaqMan Real-Time PCR ๋ถ„์„
    • ํ”ผํŽซ ๋ฐ ํ”ผํŽซ ํŒ
    • ์‹คํ—˜์‹ค์šฉ ์›์‹ฌ๋ถ„๋ฆฌ๊ธฐ
    • ์ดˆ์ €์˜จ ๋ƒ‰๋™๊ณ 
    • ๋ถ„๊ด‘ํ•™
    • ๋น„์ด์ปค
    • PCR ์žฅ๋น„ ๋ฐ ์šฉํ’ˆ
    • ์ œํ’ˆ ์นดํ…Œ๊ณ ๋ฆฌ ์ „์ฒด๋ณด๊ธฐ
  • ์‘์šฉ ๋ถ„์•ผ ๋ฐ ๊ธฐ๋ฒ•
    • ์„ธํฌ ๋ถ„์„
    • ์‹คํ—˜์‹ค ์žฅ๋น„
    • Real-Time PCR
    • PCR
    • ํฌ๋กœ๋งˆํ† ๊ทธ๋ž˜ํ”ผ
    • ์„ธํฌ ๋ฐฐ์–‘ ๋ฐ ํŠธ๋žœ์ŠคํŽ™์…˜
    • DNA/RNA ์ถ”์ถœ๊ณผ ๋ถ„์„
    • ๋‹จ๋ฐฑ์งˆ ์ƒ๋ฌผํ•™
    • ์œ ์„ธํฌ๋ถ„์„
    • ํ™”ํ•™ ์ œํ’ˆ
    • ์‘์šฉ ๋ถ„์•ผ ๋ฐ ๊ธฐ๋ฒ• ์ „์ฒด๋ณด๊ธฐ
  • ์„œ๋น„์Šค
    • ๋งž์ถคํ˜• ์„œ๋น„์Šค
    • ์—”ํ„ฐํ”„๋ผ์ด์ฆˆ๊ธ‰ ์‹คํ—˜์‹ค ์ •๋ณด ์ฒ˜๋ฆฌ
    • 360ยฐ CDMO ๋ฐ CRO ์„œ๋น„์Šค
    • CDMO ์„œ๋น„์Šค
    • ์ž„์ƒ์‹œํ—˜ CRO ์„œ๋น„์Šค
    • ์žฅ๋น„ ์„œ๋น„์Šค
    • ๊ต์œก ์„œ๋น„์Šค
    • Unity Lab Services
    • ์„œ๋น„์Šค ์ „์ฒด ๋ณด๊ธฐ
  • ์ง€์›
    • ๊ณ ๊ฐ์„ผํ„ฐ
    • ๋ฌธ์˜ํ•˜๊ธฐ
    • ์‹œํ—˜์„ฑ์ ์„œ(COA) ๋ฐ ์ ํ•ฉ์„ฑ ์ธ์ฆ์„œ(COC)
    • Safety Data Sheets (SDS)
    • ๋งค๋‰ด์–ผ
    • ์ธ์šฉ ๋ฐ ์ฐธ๊ณ  ๋ฌธํ—Œ
    • ๊ธฐ๊ธฐ ์ง€์›
    • ๊ธฐ์ˆ  ์ž๋ฃŒ/์ œํ’ˆ FAQ
    • ํ•™์Šต ์„ผํ„ฐ
    • ์ง€์› ๋ฉ”๋‰ด ์ „์ฒด๋ณด๊ธฐ
      Cell Culture Plastics
  • ์‹ ์ œํ’ˆ
  • ๋ฌธ์˜ํ•˜๊ธฐ
  • ๋น ๋ฅธ ์ฃผ๋ฌธ
  • ์ฃผ๋ฌธ ํ˜„ํ™ฉ
  • ์ œํ’ˆ ๋ฌธ์„œ ๊ฒ€์ƒ‰
Thermo Fisher Scientific Logo
Search
์ „์ฒด๊ฒ€์ƒ‰
          • ์ฃผ๋ฌธ ํ˜„ํ™ฉ
          • ๋น ๋ฅธ์ฃผ๋ฌธ
          • ๋กœ๊ทธ์ธ
            ๋กœ๊ทธ์ธ
            ํšŒ์›์ด ์•„๋‹ˆ์‹ ๊ฐ€์š”? ๊ณ„์ • ์ƒ์„ฑํ•˜๊ธฐ
            • ๊ณ„์ •์ •๋ณด
            • ์ฃผ๋ฌธ๋‚ด์—ญ์กฐํšŒ
            • ์ปค์Šคํ…€์ œํ’ˆ ๋ฐ ํ”„๋กœ์ ํŠธ
            • Services Central
          • Primary Antibodies ›
          • CD24 Antibodies
          Invitrogen
          CD24 Monoclonal Antibody (SN3), PE-DyLight™ 594
          View all (107) CD24 antibodies
          Tech Support
          Cite CD24 Monoclonal Antibody (SN3), PE-DyLight™ 594
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          CD24 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD24 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          CD24 Antibody (MA575047) in Flow
          Flow cytometry analysis of CD24 in human CD24 positive lymphocytes (red-filled); monocytes (black-dashed). Samples were incubated with monoclonal CD24 (Product # MA575047) at a dilution of 4 ฮผL reagent/100 ฮผL of peripheral whole blood, followed by PE-DyLight 594. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD24 Antibody in Flow Cytometry (Flow)
          CD24 Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as โ€œraw-uneditedโ€, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          CD24 Monoclonal Antibody (SN3), PE-DyLight™ 594
          Product Details
          MA575047
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          4 ยตL/10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          SN3
          Immunogen
          Glycoproteins purified from human NALM-1 cell line.
          3D Epitope / Immunogen
          Conjugate
          PE-DyLightโ„ข 594 PE-DyLightโ„ข 594 PE-DyLightโ„ข 594
          • Alexa Fluor 700ย 
          • APCย 
          • APC-Cyanine7
          • Biotinย 
          • FITC
          • Pacific Blue
          • PEย 
          • PerCP-Cyanine5.5
          • Request custom conjugation
          Excitation/Emission Max
          565/618 nm
          Form
          Liquid
          Purification
          Size-exclusion chromatography
          Storage buffer
          PBS, pH 7.4, with 0.2% BSA
          Contains
          15mM sodium azide
          Storage conditions
          4ยฐC, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          CD24 is a 35-50 kDa glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed on a variety of cell types, including erythrocytes, thymocytes, peripheral lymphocytes, and cells of the myeloid lineage. It is variably glycosylated, resulting in heterogeneity in molecular mass across different cell lineages, which can affect antibody staining levels on lymphocyte populations. In the context of B-cell development, CD24 is expressed from the pro-B-cell stage in the bone marrow through to mature, surface Ig-positive B cells, with very low or negative expression on plasma cells. It is also present on the majority of B-lineage acute lymphoblastic leukemias, B-cell chronic lymphocytic leukemias (CCLs), and B-cell non-Hodgkin's lymphomas. CD24 is thought to play a role in regulating B-cell proliferation and maturation, as well as in controlling autoimmunity. Additionally, CD24 has been reported to interact with P-selectin (CD62P), indicating a potential role in cell adhesion and signaling. CD24 is a valuable marker for studying B-cell development, leukemia, lymphoma, and autoimmune regulation, as well as its interactions with other molecules like P-selectin.
          Bioinformatics
          Protein Aliases: CD24 antigen (small cell lung carcinoma cluster 4 antigen); FLJ22950; FLJ43543; MGC75043
          View more View less
          Gene Aliases: CD24A
          View more View less
          UniProt ID: (Human) P25063
          View more View less
          Entrez Gene ID: (Human) 100133941
          View more View less
          Function(s)
          protein binding protein kinase binding carbohydrate binding protein tyrosine kinase activator activity Hsp70 protein binding Hsp90 protein binding
          Process(es)
          response to hypoxia cell activation regulation of cytokine-mediated signaling pathway response to molecule of bacterial origin immune response-regulating cell surface receptor signaling pathway cell adhesion positive regulation of cytosolic calcium ion concentration Wnt signaling pathway cell migration regulation of cell-cell adhesion regulation of epithelial cell differentiation T cell costimulation B cell receptor transport into membrane raft chemokine receptor transport out of membrane raft negative regulation of interleukin-6 production negative regulation of tumor necrosis factor production negative regulation of transforming growth factor beta3 production positive regulation of activated T cell proliferation cholesterol homeostasis regulation of MAPK cascade response to estrogen respiratory burst negative regulation of monocyte chemotactic protein-1 production glomerular visceral epithelial cell differentiation glomerular parietal epithelial cell differentiation intrinsic apoptotic signaling pathway cell-cell adhesion negative regulation of inflammatory response to wounding positive regulation of inflammatory response to wounding positive regulation of nephron tubule epithelial cell differentiation
          Contact tech support
          ์˜จ๋ผ์ธ ์ฃผ๋ฌธ Plus Icon Minus Icon
          • ์ฃผ๋ฌธ ํ˜„ํ™ฉ
          • ์ฃผ๋ฌธ ์ง€์›
          • ๋น ๋ฅธ ์ฃผ๋ฌธ
          • Supply Center
          • eProcurement
          ์ง€์› Plus Icon Minus Icon
          • Help and Support
          • ๊ณ ๊ฐ ์„ผํ„ฐ
          • ๊ธฐ์ˆ  ์ง€์› ์„ผํ„ฐ
          • ์ œํ’ˆ ๋ฌธ์„œ ๊ฒ€์ƒ‰
          • ์‚ฌ์ดํŠธ ๋ฌธ์ œ ๋ณด๊ณ 
          ๊ต์œก ๋ฐ ์ด๋ฒคํŠธ Plus Icon Minus Icon
          • ๊ต์œก ์„ผํ„ฐ
          • ํ”„๋กœ๋ชจ์…˜
          • ์ด๋ฒคํŠธ ๋ฐ ์›จ๋น„๋‚˜
          • ์†Œ์…œ ๋ฏธ๋””์–ด
          • ์†Œ๊ฐœ ์†Œ๊ฐœ
          • ์ฑ„์šฉ ์ฑ„์šฉ
          • ํˆฌ์ž์ž ํˆฌ์ž์ž
          • ๋‰ด์Šค ๋‰ด์Šค
          • ์‚ฌํšŒ์  ์ฑ…์ž„ ์‚ฌํšŒ์  ์ฑ…์ž„
          • Trademarks
          • ๊ณต์ •๊ฑฐ๋ž˜ ๊ณต์ •๊ฑฐ๋ž˜
          • ์ •์ฑ… ๋ฐ ๊ณ ์ง€
          • PPD
          • ์ด์šฉ ์•ฝ๊ด€
          • ๊ฐœ์ธ์ •๋ณด์ฒ˜๋ฆฌ๋ฐฉ์นจ
          • ๊ฐ€๊ฒฉ ๋ฐ ์šด์ž„ ์ •์ฑ…
          ยฉ 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea
          ๊ณ ๊ฐ์„ผํ„ฐ ๋ฌธ์˜ | ํ‰์ผ 09:00~18:00
          1661-9555   |   Live Chat   |   ์นด์นด์˜คํ†ก ์ƒ๋‹ด
          ์žฅ๋น„ ์„œ๋น„์Šค ๋ฌธ์˜ | ํ‰์ผ 09:00~18:00
          1661-5055   |   Live Chat
          ๊ณ ๊ฐ์„ผํ„ฐ ๋ฌธ์˜ | ํ‰์ผ 09:00~18:00
          1661-9555   |   Live Chat   |   ์นด์นด์˜คํ†ก ์ƒ๋‹ด
          ์žฅ๋น„ ์„œ๋น„์Šค ๋ฌธ์˜ | ํ‰์ผ 09:00~18:00
          1661-5055   |   Live Chat
          ๊ณ ๊ฐ์„ผํ„ฐ ๋ฌธ์˜ | ํ‰์ผ 09:00~18:00
          1661-9555   |   Live Chat   |   ์นด์นด์˜คํ†ก ์ƒ๋‹ด
          ์žฅ๋น„ ์„œ๋น„์Šค ๋ฌธ์˜ | ํ‰์ผ 09:00~18:00
          1661-5055   |   Live Chat
          ์จ๋ชจ ํ”ผ์…” ์‚ฌ์ด์–ธํ‹ฐํ”ฝ ์ฝ”๋ฆฌ์•„ ์ฃผ์‹ํšŒ์‚ฌ
          ๋Œ€ํ‘œ์ž : ์„์ˆ˜์ง„
          ์‚ฌ์—…์ž ๋“ฑ๋ก๋ฒˆํ˜ธ : 117-81-46910
          ์ž…๊ธˆ๊ณ„์ขŒ : ํ•˜๋‚˜์€ํ–‰ 336-890014-06204
          (์˜ˆ๊ธˆ์ฃผ : ์จ๋ชจํ”ผ์…”์‚ฌ์ด์–ธํ‹ฐํ”ฝ์ฝ”๋ฆฌ์•„ ์ฃผ์‹ํšŒ์‚ฌ)
          ์จ๋ชจ ํ”ผ์…” ์‚ฌ์ด์–ธํ‹ฐํ”ฝ ์†”๋ฃจ์…˜์Šค ์œ ํ•œํšŒ์‚ฌ
          ๋Œ€ํ‘œ์ž : ์„์ˆ˜์ง„
          ์‚ฌ์—…์ž ๋“ฑ๋ก๋ฒˆํ˜ธ : 114-86-04783
          ์ž…๊ธˆ๊ณ„์ขŒ : ์‹ ํ•œ์€ํ–‰ 140-004-396660
          (์˜ˆ๊ธˆ์ฃผ : ์จ๋ชจํ”ผ์…”์‚ฌ์ด์–ธํ‹ฐํ”ฝ์†”๋ฃจ์…˜์Šค ์œ ํ•œํšŒ์‚ฌ)
          ์ฃผ์†Œ : ์„œ์šธ์‹œ ๊ฐ•๋‚จ๊ตฌ ๊ด‘ํ‰๋กœ 281 ์ˆ˜์„œ์˜คํ”ผ์Šค๋นŒ๋”ฉ 12์ธต 06349 | ํ†ต์‹ ํŒ๋งค์—…์‹ ๊ณ ๋ฒˆํ˜ธ : 2015-์„œ์šธ๊ฐ•๋‚จ-00898
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-green-7bf6cff7cf-d8hkz:80/100.66.77.44:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0