    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD19 Antibodies
          Invitrogen
          CD19 Monoclonal Antibody (4G7), PE
          View all (389) CD19 antibodies
          Tech Support
          Cite CD19 Monoclonal Antibody (4G7), PE
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          CD19 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD19 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          CD19 Antibody (MA575114) in Flow
          Flow cytometry analysis of CD19 in human CD19 positive lymphocytes (red-filled) from neutrophil granulocytes (black-dashed). Samples were incubated with monoclonal CD19 (Product # MA575114) using a dilution of 20 µL reagent/100 µL of peripheral whole blood, followed by PE. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD19 Antibody in Flow Cytometry (Flow)
          CD19 Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          CD19 Monoclonal Antibody (4G7), PE
          Product Details
          MA575114
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          20 µL/10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          4G7
          Immunogen
          Human CCL (chronic lymphocytic leukemia) cells.
          3D Epitope / Immunogen
          Conjugate
          PE PE PE
          • Unconjugated
          • Alexa Fluor 647 
          • Alexa Fluor 700 
          • Biotin 
          • Pacific Orange
          • PE-DyLight 594
          • Request custom conjugation
          Excitation/Emission Max
          565/576 nm View spectra spectra
          Form
          Liquid
          Purification
          Size-exclusion chromatography
          Storage buffer
          PBS, pH 7.4, with 0.2% BSA
          Contains
          15mM sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          CD19 is a member of the immunoglobulin superfamily, characterized by two Ig-like domains, and is expressed on B cells throughout all stages of development, excluding terminally differentiated plasma cells. It is also expressed on follicular dendritic cells and has been observed on myeloid leukemia cells, particularly those of monocytic lineage. CD19 is considered the earliest and broadest B cell-restricted antigen, and its expression is found in all B cell precursor leukemias. CD19 forms a multimolecular complex with CD21, CD81, Leu13, MHC class II, and the B cell receptor (BCR), playing a crucial role in B cell signaling. As a signal-amplifying coreceptor for the BCR, CD19 lowers the threshold for antigen receptor-dependent stimulation, allowing B cells to respond specifically and sensitively to various antigens through low-affinity antigen receptors. Signaling through CD19 induces tyrosine phosphorylation, calcium flux, and proliferation of B cells. Beyond its role as a BCR coreceptor, CD19 can also signal independently of BCR co-ligation, serving as a central regulatory component upon which multiple signaling pathways converge. This makes CD19 an important functional regulator of both normal and malignant B cell proliferation. Mutations in the CD19 gene can result in hypogammaglobulinemia, a condition characterized by low levels of immunoglobulins, while CD19 overexpression can lead to B cell hyperactivity. CD19 is expressed on 100% of peripheral B cells, as defined by the expression of kappa or lambda light chains, underscoring its significance in B cell function and immune regulation.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: B-lymphocyte surface antigen B4; CD19 differentiation antigen; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12; unnamed protein product
          View more View less
          Gene Aliases: B4; CVID3
          View more View less
          UniProt ID: (Human) P15391
          View more View less
          Entrez Gene ID: (Human) 930
          View more View less
          Function(s)
          coreceptor activity
          Process(es)
          B cell proliferation involved in immune response immunoglobulin mediated immune response B cell proliferation antigen receptor-mediated signaling pathway B cell receptor signaling pathway regulation of B cell receptor signaling pathway regulation of B cell activation positive regulation of release of sequestered calcium ion into cytosol positive regulation of protein kinase B signaling
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-5799cd5976-hpwvt:80/100.66.77.38:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0