    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Blood Group ABH Antigen Antibodies
          Invitrogen
          Blood Group ABH Antigen Monoclonal Antibody (HE-10), PE
          View all (4) Blood Group ABH Antigen antibodies
          Questions & Answers
          Cite Blood Group ABH Antigen Monoclonal Antibody (HE-10), PE
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          Blood Group ABH Antigen Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          Blood Group ABH Antigen Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          Blood Group ABH Antigen Antibody (MA575245) in Flow
          Flow cytometry analysis of Blood Group ABH Antigen in erythrocytes (red); ABH PE concentration, black-dashed). Samples were incubated with monoclonal Blood Group ABH Antigen (Product # MA575245) using a dilution of 5 μg/mL (red), followed by PE. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Blood Group ABH Antigen Antibody in Flow Cytometry (Flow)
          Blood Group ABH Antigen Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          MA575245
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          2-5 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgM
          Class
          Monoclonal
          Type
          Antibody
          Clone
          HE-10
          Immunogen
          Mixture of erythrocytes of group A1 and glycoprotein fraction isolated from saliva of secretors with blood group A.
          Conjugate
          PE PE PE
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          565/576 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Amount
          100 µg
          Purification
          Size-exclusion chromatography
          Storage buffer
          TBS, pH 8, with 0.2% BSA
          Contains
          15mM sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          Blood-group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during malignant cellular transformation. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: ABH; BG-H; BG-H1
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch