Search
Search
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
The protein encoded by this gene is a type I membrane protein and member of the immunoglobulin superfamily. It is thought to be involved in the process of myelination. It is a lectin that binds to sialylated glycoconjugates and mediates certain myelin-neuron cell-cell interactions. Two alternatively spliced transcripts encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain neuron cytoplasmic protein 3; brain-specific neuronal polypeptide 1B236; brain-specific protein; myelin-associated glycoprotein; myelin-associated glycoprotein precursor; S-MAG (C-term.); sialic acid binding Ig-like lectin 4A; sialic acid-binding Ig-like lectin 4a; sialic acid-binding immunoglobulin-like lectin 4A; siglec-4a; unnamed protein product
Gene Aliases: 1B236; GMA; S-MAG; SIGLEC-4A; SIGLEC4; SIGLEC4A; SPG75
UniProt ID: (Human) P20916, (Mouse) P20917, (Rat) P07722
Entrez Gene ID: (Human) 4099, (Mouse) 17136, (Rat) 29409
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support