Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MAP1S Antibodies

          Abnova

          MAP1S Polyclonal Antibody, MaxPab™

          View all (8) MAP1S antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite MAP1S Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          MAP1S Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MAP1S Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MAP1S Antibody (H00055201-B01P) in WB

          Western Blot analysis of MAP1S expression in transfected 293T cell line by MAP1S MaxPab polyclonal antibody. Lane 1: BPY2IP1 transfected lysate(116.49 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MAP1S Antibody in Western Blot (WB)

          Product Details

          H00055201-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MAP1S (AAH80547.1, 1 a.a. approximately 1059 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAAVAGSGAA AAPSSLLLVV GSEFGSPGLL TYVLEELERG IRSWDVDPGV CNLDEQLKVF VSRHSATFSS IVKGQRSLHH RGDNLETLVL LNPSDKSLYD ELRNLLLDPA SHKLLVLAGP CLEETGELLL QTGGFSPHHF LQVLKDREIR DILATTPPPV QPPILTITCP TFGDWAQLAP AVPGLQGALR LQLRLNPPAQ LPNSEGLCEF LEYVAESLEP PSPFELLEPP TSGGFLRLGR PCCYIFPGGL GDAAFFAVNG FTVLVNGGSN PKSSFWKLVR HLDRVDAVLV THPGADSLPG LNSLLRRKLA ERSEVAAGGG SWDDRLRRLI SPNLGVVFFN ACEAASRLAR GEDEAELALS LLAQLGITPL PLSRGPVPAK PTVLFEKMGV GRLDMYVLHP PSAGAERTLA CVCALLVWHP AGPGEKVVRV LFPGCTPPAC LLDGLVRLQH LRFLREPVVT PQDLEGPGRA ESKESVGSRD SSKREGLLAT HPRPGQERPG VARKEPARAE APRKTEKEAK TPRELKKDPK PSVSRTQPRE VRRAASSVPN LKKTNAQAAP KPRKAPSTSH SGFPPVANGP RSPPSLRCGE ASPPSAACGS PASQLVATPS LELGPIPAGE EKALELPLAA SSIPRPRTPS PESHRSPAEG SERLSLSPLR GGEAGPDASP TVTTPTVTTP SLPAEVGSPH STEVDESLSV SFEQVLPPSA PTSEAGLSLP LRGPRARRSA SPHDVDLCLV SPCEFEHRKA VPMAPAPASP GSSNDSSARS QERAGGLGAE ETPPTSVSES LPTLSDSDPV PLAPGAADSD EDTEGFGVPR HDPLPDPLKV PPPLPDPSSI CMVDPEMLPP KTARQTENVS RTRKPLARPN SRAAAPKATP VAAAKTKGLA GGDRASRPLS ARSEPSEKGG RAPLSRKSST PKTATRGPSG SASSRPGVSA TPPKSPVYLD LAYLPSGSSA HLVDEEFFQR VRALCYVISG QDQRKEEGMR AVLDALLASK QHWDRDLQVT LIPTFDSVAM HTWYAETHAR HQALGITVLG SNSMVSMQDD AFPACKVEF

          Target Information

          Microtubule-associated protein that mediates aggregation of mitochondria resulting in cell death and genomic destruction (MAGD). Plays a role in anchoring the microtubule organizing center to the centrosomes. Binds to DNA. Plays a role in apoptosis. Involved in the formation of microtubule bundles.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: BPY2-interacting protein 1; MAP-1S; Microtubule-associated protein 1S; Microtubule-associated protein 8; OK/SW-cl.100; unnamed protein product; Variable charge Y chromosome 2-interacting protein 1; VCY2-interacting protein 1

          View more View less

          Gene Aliases: BPY2IP1; C19orf5; MAP1S; MAP8; VCY2IP-1; VCY2IP1

          View more View less

          UniProt ID: (Human) Q66K74

          View more View less

          Entrez Gene ID: (Human) 55201

          View more View less

          Function(s)
          DNA binding actin binding deoxyribonuclease activity protein binding microtubule binding tubulin binding identical protein binding beta-tubulin binding actin filament binding
          Process(es)
          microtubule cytoskeleton organization microtubule bundle formation autophagy apoptotic process mitotic spindle organization nervous system development axonogenesis brain development dendrite development regulation of microtubule depolymerization microtubule anchoring at centrosome mitochondrion transport along microtubule neuron projection morphogenesis metaphase plate congression
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline