Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • TAK1 Antibodies

          Abnova

          MAP3K7 Polyclonal Antibody, MaxPab™

          View all (43) TAK1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite MAP3K7 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          MAP3K7 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MAP3K7 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MAP3K7 Antibody (H00006885-B01P) in WB

          Western Blot analysis of MAP3K7 expression in transfected 293T cell line by MAP3K7 MaxPab polyclonal antibody. Lane 1: MAP3K7 transfected lysate(63.69 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MAP3K7 Antibody in Western Blot (WB)
          MAP3K7 Polyclonal Antibody, MaxPab™

          Product Details

          H00006885-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MAP3K7 (NP_003179.1, 1 a.a. approximately 579 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSTASAASSS SSSSAGEMIE APSQVLNFEE IDYKEIEVEE VVGRGAFGVV CKAKWRAKDV AIKQIESESE RKAFIVELRQ LSRVNHPNIV KLYGACLNPV CLVMEYAEGG SLYNVLHGAE PLPYYTAAHA MSWCLQCSQG VAYLHSMQPK ALIHRDLKPP NLLLVAGGTV LKICDFGTAC DIQTHMTNNK GSAAWMAPEV FEGSNYSEKC DVFSWGIILW EVITRRKPFD EIGGPAFRIM WAVHNGTRPP LIKNLPKPIE SLMTRCWSKD PSQRPSMEEI VKIMTHLMRY FPGADEPLQY PCQYSDEGQS NSATSTGSFM DIASTNTSNK SDTNMEQVPA TNDTIKRLES KLLKNQAKQQ SESGRLSLGA SRGSSVESLP PTSEGKRMSA DMSEIEARIA ATTGNGQPRR RSIQDLTVTG TEPGQVSSRS SSPSVRMITT SGPTSEKPTR SHPWTPDDST DTNGSDNSIP MAYLTLDHQL QPLAPCPNSK ESMAVFEQHC KMAQEYMKVQ TEIALLLQRK QELVAELDQD EKDQQNTSRL VQEHKKLLDE NKSLSTYYQQ CKKQLEVIRS QQQKRQGTS

          Target Information

          TAK1 (also known as MAP3K7), a part of the serine/threonine kinase family, is a versatile protein with many signaling functions. Originally identified as a TGF- beta-activated kinase, TAK1 interacts with TGF- beta-receptor and TRAF6 to modulate TGF- beta activation of JNK and p38. It has additional roles in activation of p38 and JNK through Wnt, BMP, and activin signaling pathways as well in response to thyroid hormone, osmotic stress, endothelin, and ephrine. Through activation of von Hippel-Lindau tumor suppressor expression, TAK1 represses PDGF-B, integrin beta1 and integrin beta5, promoting proper wound healing. TAK1 also plays an essential role in IKK activation in numerous signaling pathways including IL-1, IL-6, IL-18, TNF, CD40, TLR, and RIG-I. Activation of the TAK1-IKK pathway requires TAB2/3 and ubiquitination. In this process, TAB2/3 binds to the C-terminal region of TAK1 and becomes polyubiquitinated, TAK1 is autophosphorylated at Thr178, Thr184, Thr187 and Ser192, and finally TAK1 phosphorylates IKK- beta. Additionally, TAK1 represses human telomerase reverse transcriptase suggesting a role in regulation of cell lifespan.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: M3K7; Mitogen-activated protein kinase kinase kinase 7; RP1-154G14.1; TAK1d; TGF-beta activated kinase 1; TGF-beta-activated kinase 1; Transforming growth factor-beta-activated kinase 1; unnamed protein product

          View more View less

          Gene Aliases: CSCF; FMD2; MAP3K7; MEKK7; TAK1; TGF1a

          View more View less

          UniProt ID: (Human) O43318

          View more View less

          Entrez Gene ID: (Human) 6885

          View more View less

          Function(s)
          nucleotide binding magnesium ion binding transcription coactivator binding protein kinase activity protein serine/threonine kinase activity MAP kinase activity MAP kinase kinase kinase activity receptor binding type II transforming growth factor beta receptor binding protein binding ATP binding MAP kinase kinase kinase kinase activity kinase activity transferase activity protein kinase binding receptor tyrosine kinase binding ubiquitin protein ligase binding histone kinase activity identical protein binding metal ion binding scaffold protein binding protein serine kinase activity protein serine/threonine kinase binding DNA-binding transcription factor binding linear polyubiquitin binding
          Process(es)
          MAPK cascade response to hypoxia stimulatory C-type lectin receptor signaling pathway positive regulation of T cell cytokine production cytoplasmic pattern recognition receptor signaling pathway MyD88-dependent toll-like receptor signaling pathway chromatin remodeling apoptotic process inflammatory response immune response signal transduction transforming growth factor beta receptor signaling pathway I-kappaB kinase/NF-kappaB signaling I-kappaB phosphorylation JNK cascade positive regulation of metabolic process positive regulation of autophagy negative regulation of gene expression positive regulation of macroautophagy signal transduction involved in regulation of gene expression SCF-dependent proteasomal ubiquitin-dependent protein catabolic process positive regulation of type I interferon production protein exit from endoplasmic reticulum positive regulation of interleukin-2 production toll-like receptor 3 signaling pathway toll-like receptor 4 signaling pathway TRIF-dependent toll-like receptor signaling pathway nucleotide-binding domain, leucine rich repeat containing receptor signaling pathway NIK/NF-kappaB signaling p38MAPK cascade Fc-epsilon receptor signaling pathway interleukin-33-mediated signaling pathway interleukin-17A-mediated signaling pathway defense response to bacterium signal transduction in response to DNA damage positive regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of I-kappaB kinase/NF-kappaB signaling anoikis positive regulation of MAPK cascade positive regulation of JUN kinase activity positive regulation of cell cycle positive regulation of cell size T cell receptor signaling pathway stress-activated MAPK cascade interleukin-1-mediated signaling pathway cellular response to tumor necrosis factor cellular response to hypoxia cellular response to transforming growth factor beta stimulus cGAS/STING signaling pathway positive regulation of cGAS/STING signaling pathway negative regulation of cytokine production involved in inflammatory response positive regulation of NIK/NF-kappaB signaling cellular response to angiotensin positive regulation of vascular smooth muscle cell proliferation positive regulation of vascular associated smooth muscle cell migration response to angiotensin
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline