Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • MAPKBP1 Antibodies

          Abnova

          MAPKBP1 Polyclonal Antibody, MaxPab™

          View all (7) MAPKBP1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite MAPKBP1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          MAPKBP1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MAPKBP1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MAPKBP1 Antibody (H00023005-B01P) in WB

          Western Blot analysis of MAPKBP1 expression in transfected 293T cell line by MAPKBP1 MaxPab polyclonal antibody. Lane 1: MAPKBP1 transfected lysate(111.65 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MAPKBP1 Antibody in Western Blot (WB)

          Product Details

          H00023005-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MAPKBP1 (AAH36660.1, 1 a.a. approximately 1015 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MGTLRVHELQ SLSEMLKVEA HDSEILCLEY SKPDTGLKLL ASASRDRLIH VLDAGREYSL QQTLDEHSSS ITAVKFAASD GQVRMISCGA DKSIYFRTAQ KSGDGVQFTR THHVVRKTTL YDMDVEPSWK YTAIGCQDRN IRIFNISSGK QKKLFKGSQG EDGTLIKVQT DPSGIYIATS CSDKNLSIFD FSSGECVATM FGHSEIVTGM KFSNDCKHLI SVSGDSCIFV WRLSSEMTIS MRQRLAELRQ RQRGGKQQGP SSPQRASGPN RHQAPSMLSP GPALSSDSDK EGEDEGTEEE LPALPVLAKS TKKALASVPS PALPRSLSHW EMSRAQESVG FLDSAPAANP GPRRRGRWVQ PGVELSVRSM LDLRQLETLA PSLQDPSQDS LAIIPSGPRK HGQEALETSL TSQNEKPPRP QASQPCSYPH IIRLLSQEEG VFAQDLEPAP IEDGIVYPEP SDNPTMDTSE FQVQAPARGT LGRVYPGSRS SEKHSPDSAC SVDYSSSCLS SPEHPTEDSE STEPLSVDGI SSDLEEPAEG DEEEEEEEGG MGPYGLQEGS PQTPDQEQFL KQHFETLASG AAPGAPVQVP ERSESRSISS RFLLQVQTRP LREPSPSSSS LALMSRPAQV PQASGEQPRG NGANPPGAPP EVEPSSGNPS PQQAASVLLP RCRLNPDSSW APKRVATASP FSGLQKAQSV HSLVPQERHE ASLQAPSPGA LLSREIEAQD GLGSLPPADG PPSRPHSYQN PTTSSMAKIS RSISVGENLG LVAEPQAHAP IRVSPLSKLA LPSRAHLVLD IPKPLPDRPT LAAFSPVTKG RAPGEAEKPG FPVGLGKAHS TTERWACLGE GTTPKPRTEC QAHPGPSSPC AQQLPVSSLF QGPENLQPPP PEKTPNPMEC TKPGAALSQD SEPAVSLEQC EQLVAELRGS VRQAVRLYHS VAGCKMPSAE QSRIAQLLRD TFSSVRQELE AVAGAVLSSP GSSPGAVGAE QTQALLEQYS ELLLRAVERR MERKL

          Target Information

          MAP kinases play a significant role in many biological processes, including cell adhesion and spreading, cell differentiation and apoptosis. MAPKBP-1 (mitogen-activated protein kinase binding protein 1), also known as JNKBP-1, is a 1,514 amino acid protein that contains twelve WD repeats. Induced by TRAF2 (TNF receptor-associated factor 2) and Tak1 (TGF-beta-activated kinase 1), MAPKBP-1 is thought to act an adaptor protein for NFkappaB (nuclear factor kappa-B) activation. MAPKBP-1 interacts with JNK3 and may promote TRAF2 polyubiquitination. MAPKBP-1 exists as six alternatively spliced variants and is encoded by a gene located on human chromosome 15. Human chromosome 15 houses over 700 genes and comprises nearly 3% of the human genome. Angelman syndrome, Prader-Willi syndrome, Tay-Sachs disease and Marfan syndrome are all associated with defects in chromosome 15-localized genes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: JNK-binding protein 1; JNKBP-1; Mitogen-activated protein kinase-binding protein 1; unnamed protein product

          View more View less

          Gene Aliases: JNKBP-1; JNKBP1; KIAA0596; MAPKBP1; NPHP20

          View more View less

          UniProt ID: (Human) O60336

          View more View less

          Entrez Gene ID: (Human) 23005

          View more View less

          Function(s)
          protein binding
          Process(es)
          negative regulation of interleukin-8 production negative regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of JNK cascade negative regulation of defense response to bacterium regulation of intracellular signal transduction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline