Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • MCF2L Antibodies

          Abnova

          MCF2L Polyclonal Antibody, MaxPab™

          View all (5) MCF2L antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite MCF2L Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          MCF2L Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MCF2L Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MCF2L Antibody (H00023263-B01P) in WB

          Western Blot analysis of MCF2L expression in transfected 293T cell line by MCF2L MaxPab polyclonal antibody. Lane 1: MCF2L transfected lysate(108.24 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MCF2L Antibody in Western Blot (WB)
          MCF2L Polyclonal Antibody, MaxPab™

          Product Details

          H00023263-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MCF2L (NP_079255, 1 a.a. approximately 984 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MTVRRLSLLC RDLWALWLLL KAGADEIMHQ DIVPLCAADI QDQLKKRFAY LSGGRGQDGS PVITFPDYPA FSEIPDKEFQ NVMTYLTSIP SLQDAGIGFI LVIDRRRDKW TSVKASVLRI AASFPANLQL VLVLRPTGFF QRTLSDIAFK FNRDDFKMKV PVIMLSSVPD LHGYIDKSQL TEDLGGTLDY CHSRWLCQRT AIESFALMVK QTAQMLQSFG TELAETELPN DVQSTSSVLC AHTEKKDKAK EDLRLALKEG HSVLESLREL QAEGSEPSVN QDQLDNQATV QRLLAQLNET EAAFDEFWAK HQQKLEQCLQ LRHFEQGFRE VKAILDAASQ KIATFTDIGN SLAHVEHLLR DLASFEEKSG VAVERARALS LDGEQLIGNK HYAVDSIRPK CQELRHLCDQ FSAEIARRRG LLSKSLELHR RLETSMKWCD EGIYLLASQP VDKCQSQDGA EAALQEIEKF LETGAENKIQ ELNAIYKEYE SILNQDLMEH VRKVFQKQAS MEEVFHRRQA SLKKLAARQT RPVQPVAPRP EALAKSPCPS PGIRRGSENS SSEGGALRRG PYRRAKSEMS ESRQGRGSAG EEEESLAILR RHVMSELLDT ERAYVEELLC VLEGYAAEMD NPLMAHLLST GLHNKKDVLF GNMEEIYHFH NRIFLRELEN YTDCPELVGR CFLERMEDFQ IYEKYCQNKP RSESLWRQCS DCPFFQECQR KLDHKLSLDS YLLKPVQRIT KYQLLLKEML KYSRNCEGAE DLQEALSSIL GILKAVNDSM HLIAITGYDG NLGDLGKLLM QGSFSVWTDH KRGHTKVKEL ARFKPMQRHL FLHEKAVLFC KKREENGEGY EKAPSYSYKQ SLNMAAVGIT ENVKGDAKKF EIWYNAREEV YIVQAPTPEI KAAWVNEIRK VLTSQLQACR EASQHRALEQ SQSLPLPAPT STSPSRGNSR NIKKLEERKT DPLSLEGYVS SAPLTKPPEK GKEP

          Target Information

          This gene encodes a guanine nucleotide exchange factor that interacts specifically with the GTP-bound Rac1 and plays a role in the Rho/Rac signaling pathways. A variant in this gene was associated with osteoarthritis. Alternative splicing results in multiple transcript variants.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: DBL's big sister; FLJ12122; Guanine nucleotide exchange factor DBS; KIAA0362; MCF2 transforming sequence-like protein; MCF2-transforming sequence-like protein; RP11-98F14.2; unnamed protein product

          View more View less

          Gene Aliases: ARHGEF14; DBS; KIAA0362; MCF2L; OST

          View more View less

          UniProt ID: (Human) O15068

          View more View less

          Entrez Gene ID: (Human) 23263

          View more View less

          Function(s)
          guanyl-nucleotide exchange factor activity protein binding lipid binding phosphatidylinositol binding
          Process(es)
          positive regulation of Rho protein signal transduction intracellular signal transduction regulation of small GTPase mediated signal transduction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline