Search
Search
Active Motif
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
N6-methylated adenine (m6A) is prevalently present in nearly all RNA types and can be found in all organisms from bacteria to humans. It preferentially appears around stop codons and within long internal exons in mammalian messager RNAs. m6A plays an important role in the efficiency of mRNA splicing, processing, translation efficiency, editing and mRNA stability. m6A also takes place in other RNA molecules, such as primary miRNA (pri-miRNAs). METTL14 (methyltransferase-like 14) forms a stable N6 methyltransferase heterodimer complex with METTL3, which catalyzes the generation of m6A modification on mammalian nuclear RNAs. METTL3 is the catalytically active subunit, while METTL14 plays a structural role critical for substrate recognition.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D1075-like; D1075-like gene product; methyltransferase like 1; methyltransferase-like protein 1; miRNA (guanine-N(7)-)-methyltransferase; mRNA (guanine-N(7)-)-methyltransferase; protein Wuho homolog; TRM82 tRNA methyltransferase 82 homolog; tRNA (guanine(46)-N(7))-methyltransferase; tRNA (guanine-N(7)-)-methyltransferase subunit WDR4; tRNA(m7G46)-methyltransferase; unnamed protein product; WD repeat domain 4; WD repeat domain 4 METTL1 scaffold protein; WD repeat-containing protein 4
Gene Aliases: C12orf1; GAMOS6; hWH; MIGSB; TRM8; TRM82; TRMT8; TRMT82; Wuho; YDL201w
UniProt ID: (Human) Q9UBP6, (Human) P57081
Entrez Gene ID: (Human) 4234, (Human) 10785
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support