Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Mgea5 Antibodies

          Proteintech

          MGEA5 Polyclonal Antibody

          66 References
          View all (18) Mgea5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite MGEA5 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (22)
          MGEA5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MGEA5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 22

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MGEA5 Antibody (14711-1-AP) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using MGEA5 antibody (Product # 14711-1-AP) at dilution of 1:200 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L), CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MGEA5 Antibody in Immunocytochemistry (ICC/IF)
          MGEA5 Antibody in Immunocytochemistry (ICC/IF)
          MGEA5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MGEA5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MGEA5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MGEA5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MGEA5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MGEA5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MGEA5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MGEA5 Antibody in Western Blot (WB)
          MGEA5 Antibody in Western Blot (WB)
          MGEA5 Antibody in Western Blot (WB)
          MGEA5 Antibody in Western Blot (WB)
          MGEA5 Antibody in Western Blot (WB)
          MGEA5 Antibody in Western Blot (WB)
          MGEA5 Antibody in Western Blot (WB)
          MGEA5 Antibody in Western Blot (WB)
          MGEA5 Antibody in Western Blot (WB)
          MGEA5 Antibody in Western Blot (WB)
          MGEA5 Antibody in Western Blot (WB)
          MGEA5 Antibody in Immunoprecipitation (IP)
          MGEA5 Antibody in Immunoprecipitation (IP)
          MGEA5 Polyclonal Antibody

          Product Details

          14711-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          View 43 publications 43 publications

          Immunohistochemistry (IHC)

          -
          View 8 publications 8 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 7 publications 7 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication

          ChIP assay (ChIP)

          Assay-dependent
          View 1 publication 1 publication

          RNA Immunoprecipitation (RIP)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 5 publications 5 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Fruit fly, Human, Mouse, Pig, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MGEA5 Fusion Protein Ag6405 (1-350 aa encoded by BC039583)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          30 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Two bands at 130 kDa and 75 kDa could be detected using antibody 66033-1-Ig.

          Immunogen sequence: MVQKESQAT LEERESELSS NPAASAGASL EPPAAPAPGE DNPAGAGGAA VAGAAGGARR FLCGVVEGFY GRPWVMEQRK ELFRRLQKWE LNTYLYAPKD DYKHRMFWRE MYSVEEAEQL MTLISAAREY EIEFIYAISP GLDITFSNPK EVSTLKRKLD QVSQFGCRSF ALLFDDIDHN MCAADKEVFS SFAHAQVSIT NEIYQYLGEP ETFLFCPTEY CGTFCYPNVS QSPYLRTVGE KLLPGIEVLW TGPKVVSKEI PVESIEEVSK IIKRAPVIWD NIHANDYDQK RLFLGPYKGR STELIPRLKG VLTNPNCEFE ANYVAIHTLA TWYKSNMNGV RKDVVMSRQV A (1-350 aa encoded by BC039583)

          Target Information

          This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This protein forms homo- or hetero-dimers with other Ikaros family members, and is thought to function predominantly in early hematopoietic development. Multiple transcript variants encoding different isoforms have been found for this gene, but the biological validity of some variants has not been determined.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Beta-hexosaminidase; Beta-N-acetylglucosaminidase; Beta-N-acetylhexosaminidase; Bifunctional protein NCOAT; hyaluronidase in meningioma; meningioma expressed antigen 5 (hyaluronidase); Meningioma-expressed antigen 5; MGEA5; N-acetyl-beta-D-glucosaminidase; N-acetyl-beta-glucosaminidase; NCOAT; Nuclear cytoplasmic O-GlcNAcase and acetyltransferase; OGA; Protein O-GlcNAcase; unnamed protein product

          View more View less

          Gene Aliases: 2810009A20Rik; 4833427O07Rik; 5830447M11Rik; AA408215; HEXC; Hy5; KIAA0679; MEA5; MGEA5; mKIAA0679; NCOAT; O-GLCNACASE; OGA

          View more View less

          UniProt ID: (Rat) Q8VIJ5, (Mouse) Q9EQQ9, (Human) O60502

          View more View less

          Entrez Gene ID: (Rat) 154968, (Mouse) 76055, (Human) 10724

          View more View less

          Function(s)
          histone acetyltransferase activity beta-N-acetylglucosaminidase activity beta-N-acetylhexosaminidase activity hydrolase activity hydrolase activity, acting on glycosyl bonds hyalurononglucosaminidase activity hexosaminidase activity identical protein binding [protein]-3-O-(N-acetyl-D-glucosaminyl)-L-serine/L-threonine O-N-acetyl-alpha-D-glucosaminase activity catalytic activity, acting on a protein hydrolase
          Process(es)
          N-acetylglucosamine metabolic process protein deglycosylation protein targeting to membrane aging positive regulation of calcium ion transport into cytosol negative regulation of cardiac muscle adaptation histone acetylation positive regulation of cell killing positive regulation of insulin secretion positive regulation of protein complex disassembly positive regulation of proteolysis dATP metabolic process positive regulation of glucose import response to steroid hormone positive regulation of DNA metabolic process positive regulation of mitochondrial depolarization positive regulation of calcium ion transport negative regulation of protein glycosylation positive regulation of growth hormone secretion necrotic cell death metabolic process protein O-linked glycosylation glycoprotein catabolic process glycoprotein metabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-548c9458fd-dmg5x:80/100.66.73.23:80.
          git-commit: b50e517baec6e51dce58f17cb59caa961a96861c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.53.0-Offline