Search
Search
OriGene
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
MIF4GD is a protein which contains an MIF4G domain.This gene encodes a protein which contains an MIF4G domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hSLIP1; MIF4G domain-containing protein; SLBP (stem loop binding protein)-interacting protein 1; SLBP-interacting protein 1; unnamed protein product
Gene Aliases: AD023; MIF4GD; MIFD; SLIP1
UniProt ID: (Human) A9UHW6
Entrez Gene ID: (Human) 57409
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support