Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • MKKS Antibodies

          Abnova

          MKKS Polyclonal Antibody, MaxPab™

          View all (5) MKKS antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite MKKS Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          MKKS Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MKKS Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MKKS Antibody (H00008195-D01P) in WB

          MKKS MaxPab rabbit polyclonal antibody. Western Blot analysis of MKKS expression in A-431. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MKKS Antibody in Western Blot (WB)
          MKKS Polyclonal Antibody, MaxPab™

          Product Details

          H00008195-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MKKS (NP_061336.1, 1 a.a. approximately 570 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSRLEAKKPS LCKSEPLTTE RVRTTLSVLK RIVTSCYGPS GRLKQLHNGF GGYVCTTSQS SALLSHLLVT HPILKILTAS IQNHVSSFSD CGLFTAILCC NLIENVQRLG LTPTTVIRLN KHLLSLCISY LKSETCGCRI PVDFSSTQIL LCLVRSILTS KPACMLTRKE TEHVSALILR AFLLTIPENA EGHIILGKSL IVPLKGQRVI DSTVLPGILI EMSEVQLMRL LPIKKSTALK VALFCTTLSG DTSDTGEGTV VVSYGVSLEN AVLDQLLNLG RQLISDHVDL VLCQKVIHPS LKQFLNMHRI IAIDRIGVTL MEPLTKMTGT QPIGSLGSIC PNSYGSVKDV CTAKFGSKHF FHLIPNEATI CSLLLCNRND TAWDELKLTC QTALHVLQLT LKEPWALLGG GCTETHLAAY IRHKTHNDPE SILKDDECTQ TELQLIAEAF CSALESVVGS LEHDGGEILT DMKYGHLWSV QADSPCVANW PDLLSQCGCG LYNSQEELNW SFLRSTRRPF VPQSCLPHEA VGSASNLTLD CLTAKLSGLQ VAVETANLIL DLSYVIEDKN

          Target Information

          Probable molecular chaperone. Assists the folding of proteins upon ATP hydrolysis. As part of the BBS/CCT complex may play a role in the assembly of BBSome, a complex involved in ciliogenesis regulating transports vesicles to the cilia. May play a role in protein processing in limb, cardiac and reproductive system development. May play a role in cytokinesis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: alternative protein MKKS; Bardet-Biedl syndrome 6 protein; McKusick-Kaufman/Bardet-Biedl syndromes putative chaperonin; Molecular chaperone MKKS; unnamed protein product

          View more View less

          Gene Aliases: BBS6; HMCS; KMS; MKKS; MKS

          View more View less

          UniProt ID: (Human) Q9NPJ1

          View more View less

          Entrez Gene ID: (Human) 8195

          View more View less

          Function(s)
          nucleotide binding protein binding ATP binding unfolded protein binding RNA polymerase II sequence-specific DNA binding transcription factor binding
          Process(es)
          heart looping developmental process involved in reproduction protein folding spermatid development determination of left/right symmetry heart development visual perception sensory perception of smell gonad development gene expression negative regulation of gene expression artery smooth muscle contraction striatum development hippocampus development cerebral cortex development negative regulation of actin filament polymerization melanosome transport developmental process leptin-mediated signaling pathway social behavior negative regulation of appetite by leptin-mediated signaling pathway positive regulation of multicellular organism growth vasodilation response to leptin fat cell differentiation photoreceptor cell maintenance negative regulation of blood pressure intracellular transport animal organ development system development brain morphogenesis detection of mechanical stimulus involved in sensory perception of sound chaperone-mediated protein complex assembly cartilage development regulation of stress fiber assembly pigment granule aggregation in cell center convergent extension involved in gastrulation cilium morphogenesis regulation of cilium beat frequency involved in ciliary motility face development response to inositol non-motile cilium assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline