Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MTTAASEVP SFLVERMANV RRRRQDRRGM EGGILKSRIV TWEPSEEFVR NNHVINTPLQ TMHIMADLGP YKKLGYKKYE HVLCTLKVDS NGVITVKPDF TGLKGPYRIE TEGEKQELWK YTIDNVSPHA QPEEEERERR VFKDLYGRHK EYLSSLVGTD FEMTVPGALR LFVNGEVVSA QGYEYDNLYV HFFVELPTAH WSSPAFQQLS GVTQTCTTKS LAMDKVAHFS YPFTFEAFFL HEDESSDALP EWPVLYCEVL SLDFWQRYRV EGYGAVVLPA TPGSHTLTVS TWRPVELGTV AELRRFFIGG SLELEDLSYV RI (1-321 aa encoded by BC010061)
The protein encoded by this gene localizes to the basal body and is required for formation of the primary cilium in ciliated epithelial cells. Mutations in this gene result in Meckel syndrome type 1 and in Bardet-Biedl syndrome type 13. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20345; meckel syndrome type 1 protein; meckel syndrome type 1 protein homolog; Meckel syndrome, type 1; POC12 centriolar protein homolog; unnamed protein product
Gene Aliases: avc6; B8d3; BBS13; JBTS28; MES; MKS; POC12
UniProt ID: (Human) Q9NXB0, (Mouse) Q5SW45, (Rat) Q499Q5
Entrez Gene ID: (Human) 54903, (Mouse) 380718, (Rat) 287612
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support