Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • MYST1 Antibodies

          Proteintech

          MOF Polyclonal Antibody

          9 References
          View all (16) MYST1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite MOF Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (9)
          MOF Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MOF Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MOF Antibody (13842-1-AP) in WB

          Rat liver tissue were subjected to SDS PAGE followed by western blot with 13842-1-AP (MOF Antibody) at dilution of 1:600 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MOF Antibody in Western Blot (WB)
          MOF Antibody in Western Blot (WB)
          MOF Antibody in Western Blot (WB)
          MOF Antibody in Western Blot (WB)
          MOF Antibody in Western Blot (WB)
          MOF Antibody in Western Blot (WB)
          MOF Antibody in Western Blot (WB)
          MOF Antibody in Western Blot (WB)
          MOF Antibody in Immunoprecipitation (IP)

          Product Details

          13842-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 7 publications 7 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MOF Fusion Protein Ag4802 (118-467 aa encoded by BC037773)

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.19 mg/mL

          Amount

          28.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: AVQKNSEKY LSELAEQPER KITRNQKRKH DEINHVQKTY AEMDPTTAAL EKEHEAITKV KYVDKIHIGN YEIDAWYFSP FPEDYGKQPK LWLCEYCLKY MKYEKSYRFH LGQCQWRQPP GKEIYRKSNI SVYEVDGKDH KIYCQNLCLL AKLFLDHKTL YFDVEPFVFY ILTEVDRQGA HIVGYFSKEK ESPDGNNVAC ILTLPPYQRR GYGKFLIAFS YELSKLESTV GSPEKPLSDL GKLSYRSYWS WVLLEILRDF RGTLSIKDLS QMTSITQNDI ISTLQSLNMV KYWKGQHVIC VTPKLVEEHL KSAQYKKPPI TGGWGAAVCR GRWGSVSMWT GRSQGLLIAV T (118-467 aa encoded by BC037773)

          Target Information

          The MYST family of histone acetyltransferases, which includes MYST1, is named for the founding members MOZ (MYST3), yeast YBF2 and SAS2, and TIP60 (HTATIP). All members of this family contain a MYST region of about 240 amino acids with a canonical acetyl-CoA-binding site and a C2HC-type zinc finger motif. Most MYST proteins also have a chromodomain involved in protein-protein interactions and targeting transcriptional regulators to chromatin.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: h FLJ14040; Histone acetyltransferase KAT8; histone acetyltransferase MYST1; hMOF; K(lysine) acetyltransferase 8; Lysine acetyltransferase 8; Males-absent on the first protein homolog; mMof; MOF; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1; MYST family member; MYST histone acetyltransferase 1; MYST protein 1; MYST-1; ortholog of Drosophila males absent on the first (MOF); probable histone acetyltransferase MYST1; Protein acetyltransferase KAT8; Protein propionyltransferase KAT8; unnamed protein product

          View more View less

          Gene Aliases: 2010203C02Rik; 5830450F21Rik; D7Ertd629e; hMOF; KAT8; LIGOWS; MOF; MYST1; PP7073; ZC2HC8

          View more View less

          UniProt ID: (Human) Q9H7Z6, (Mouse) Q9D1P2, (Rat) Q5XI06

          View more View less

          Entrez Gene ID: (Human) 84148, (Mouse) 67773, (Rat) 310194

          View more View less

          Function(s)
          transcription coactivator activity histone acetyltransferase activity protein binding zinc ion binding H4 histone acetyltransferase activity acetyltransferase activity transferase activity transferase activity, transferring acyl groups enzyme binding histone acetyltransferase activity (H4-K5 specific) histone acetyltransferase activity (H4-K8 specific) metal ion binding histone acetyltransferase activity (H4-K16 specific) RNA polymerase II sequence-specific DNA binding transcription factor binding peptide-lysine-N-acetyltransferase activity protein propionyltransferase activity DNA-binding transcription factor binding promoter-specific chromatin binding transcription factor binding transferase activity, transferring acyl groups other than amino-acyl groups methylated histone binding molecular_function histone modifying enzyme
          Process(es)
          epithelial to mesenchymal transition chromatin organization regulation of transcription, DNA-templated dosage compensation by inactivation of X chromosome regulation of autophagy negative regulation of macromolecule biosynthetic process positive regulation of epithelial to mesenchymal transition negative regulation of epithelial to mesenchymal transition skeletal muscle satellite cell differentiation neurogenesis hemopoiesis myeloid cell differentiation negative regulation of type I interferon production positive regulation of type I interferon production post-embryonic hemopoiesis regulation of gene expression, epigenetic regulation of cell differentiation positive regulation of gene expression, epigenetic negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated oogenesis regulation of mRNA processing negative regulation of multicellular organismal process positive regulation of transcription initiation from RNA polymerase II promoter membraneless organelle assembly positive regulation of skeletal muscle satellite cell differentiation regulation of transcription from mitochondrial promoter transcription, DNA-templated chromatin modification histone acetylation histone H4-K5 acetylation histone H4-K8 acetylation histone H4-K16 acetylation biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline