Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • MPP5 Antibodies

          Abnova

          MPP5 Polyclonal Antibody, MaxPab™

          View all (12) MPP5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite MPP5 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          MPP5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MPP5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MPP5 Antibody (H00064398-D01P) in WB

          Western Blot analysis of MPP5 expression in transfected 293T cell line by MPP5 MaxPab polyclonal antibody. Lane 1: MPP5 transfected lysate(77.30 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MPP5 Antibody in Western Blot (WB)
          MPP5 Polyclonal Antibody, MaxPab™

          Product Details

          H00064398-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MPP5 (NP_071919.2, 1 a.a. approximately 675 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MTTSHMNGHV TEESDSEVKN VDLASPEEHQ KHREMAVDCP GDLGTRMMPI RRSAQLERIR QQQEDMRRRR EEEGKKQELD LNSSMRLKKL AQIPPKTGID NPMFDTEEGI VLESPHYAVK ILEIEDLFSS LKHIQHTLVD SQSQEDISLL LQLVQNKDFQ NAFKIHNAIT VHMNKASPPF PLISNAQDLA QEVQTVLKPV HHKEGQELTA LLNTPHIQAL LLAHDKVAEQ EMQLEPITDE RVYESIGQYG GETVKIVRIE KARDIPLGAT VRNEMDSVII SRIVKGGAAE KSGLLHEGDE VLEINGIEIR GKDVNEVFDL LSDMHGTLTF VLIPSQQIKP PPAKETVIHV KAHFDYDPSD DPYVPCRELG LSFQKGDILH VISQEDPNWW QAYREGDEDN QPLAGLVPGK SFQQQREAMK QTIEEDKEPE KSGKLWCAKK NKKKRKKVLY NANKNDDYDN EEILTYEEMS LYHQPANRKR PIILIGPQNC GQNELRQRLM NKEKDRFASA VPHTTRSRRD QEVAGRDYHF VSRQAFEADI AAGKFIEHGE FEKNLYGTSI DSVRQVINSG KICLLSLRTQ SLKTLRNSDL KPYIIFIAPP SQERLRALLA KEGKNPKPEE LREIIEKTRE MEQNNGHYFD TAIVNSDLDK AYQELLRLIN KLDTEPQWVP STWLR

          Target Information

          MPP5 (Membrane Protein, Palmitoylated 5) plays a critical role in neurogenesis and hematopoiesis. This protein is crucial for establishing and maintaining cell polarity in epithelial cells and neurons. It is involved in the Par-3 complex, which contributes to maintaining tight junctions in epithelial cells. Mutations in MPP5 have been associated with severe neurologic impairments, including global developmental delay (GDD) and behavioral changes. In animal models, MPP5 is highly involved in the early differentiation of multipotent progenitors and supports long-term lineage-specific contributions, primarily in myeloid and lymphoid cells. MPP5 variants disrupt normal cellular function, leading to profound developmental and functional abnormalities.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: FLJ12615; MAGUK p55 subfamily member 5; membrane palmitoylated protein 5; Membrane protein, palmitoylated 5; membrane protein, palmitoylated 5 (MAGUK p55 subfamily member 5); palmitoylated 5; protein associated with lin seven 1; Protein associated with Lin-7 1; protein associated with LIN7 1, MAGUK family member; Protein PALS1; stardust; unnamed protein product

          View more View less

          Gene Aliases: MPP5; PALS1

          View more View less

          UniProt ID: (Human) Q8N3R9

          View more View less

          Entrez Gene ID: (Human) 64398

          View more View less

          Function(s)
          nucleotide binding protein binding ATP binding protein domain specific binding identical protein binding
          Process(es)
          morphogenesis of an epithelial sheet plasma membrane organization protein localization gene expression establishment or maintenance of polarity of embryonic epithelium regulation of transforming growth factor beta receptor signaling pathway central nervous system neuron development cerebral cortex development peripheral nervous system myelin maintenance myelin assembly protein localization to myelin sheath abaxonal region establishment or maintenance of epithelial cell apical/basal polarity generation of neurons protein localization to plasma membrane
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline