Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody is a rabbit polyclonal antibody raised against the full length MRPS35 protein. It specifically reacts with the 37 kDa MRPS35protein.
Immunogen sequence: MAAAALPAW LSLQSRARTL RAFSTAVYSA TPVPTPSLPE RTPGNERPPR RKALPPRTEK MAVDQDWPSV YPVAAPFKPS AVPLPVRMGY PVKKGVPMAK EGNLELLKIP NFLHLTPVAI KKHCEALKDF CTEWPAALDS DEKCEKHFPI EIDSTDYVSS GPSVRNPRAR VVVLRVKLSS LNLDDHAKKK LIKLVGERYC KTTDVLTIKT DRCPLRRQNY DYAVYLLTVL YHESWNTEEW EKSKTEADME EYIWENSSSE RNILETLLQM KAAEKNMEIN KEELLGTKEI EEYKKSVVSL KNEEENENSI SQYKESVKRL LNVT (1-323 aa encoded by BC015862)
Mammalian mitochondrial ribosomes (mitoribosomes) are responsible for protein synthesis within the mitochondrion. The mitoribosomes are composed of a 4:1 ratio of protein to RNA, with the proteins forming two subunits, the 28S subunit and the 39S subunit. Across species, the proteins that make up the mitoribosome subunits vary greatly in sequence, preventing easy recognition by sequence homology. MRP-S35 (mitochondrial ribosomal protein S35), also known as MDS023, MRPS28 or HDCMD11P, is a 323 amino acid protein that localizes to the mitochondrion, where it exists as a component of the 28S ribosomal subunit and works in conjunction with other MRPs to mediate protein synthesis. Existing as two alternatively spliced isoforms, MRP-S35 is encoded by a gene located on human chromosome 10, which houses over 1,200 genes and comprises nearly 4. 5% of the human genome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 28S ribosomal protein S28, mitochondrial; 28S ribosomal protein S35, mitochondrial; mitochondrial ribosomal protein S28; mitochondrial ribosomal protein S34; mitochondrial small ribosomal subunit protein mS35; MRP-S35; S28mt; S35mt; unnamed protein product
Gene Aliases: HDCMD11P; MDS023; MDSO23; MRP-S28; MRPS28; mS35
UniProt ID: (Human) P82673, (Mouse) Q8BJZ4
Entrez Gene ID: (Human) 60488, (Mouse) 232536, (Rat) 297727
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support