          • Primary Antibodies ›
          • Melan-A Antibodies
          NeoBiotechnologies
          Melanoma Marker (MART-1 + Tyrosinase + gp100) Monoclonal Antibody (M2-7C10, M2-9E3, T311, HMB45)
          View all (116) Melan-A antibodies
          Tech Support
          Cite Melanoma Marker (MART-1 + Tyrosinase + gp100) Monoclonal Antibody (M2-7C10, M2-9E3, T311, HMB45)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          Melanoma Marker (MART-1 + Tyrosinase + gp100) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Melanoma Marker (MART-1 + Tyrosinase + gp100) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          Melanoma Marker (MART-1 + Tyrosinase + gp100) Antibody (MSM1-701-P0) in IHC (P)
          Formalin-fixed, paraffin-embedded human Melanoma stained with Melanoma Marker Monoclonal Antibody (M2-7C10 +M2-9E3 +T311 +HMB45). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Melanoma Marker (MART-1 + Tyrosinase + gp100) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Melanoma Marker (MART-1 + Tyrosinase + gp100) Antibody in Western Blot (WB)
          Melanoma Marker (MART-1 + Tyrosinase + gp100) Monoclonal Antibody (M2-7C10, M2-9E3, T311, HMB45)
          Product Details
          MSM1-701-P0
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          2-4 µg/mL
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          Assay-dependent
          -
          Immunohistochemistry (PFA fixed) (IHC (PFA))
          1-2 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          M2-7C10, M2-9E3, T311, HMB45
          Immunogen
          Recombinant hMART-1 protein (M2-7C10. M2-9E3). Recombinant tyrosinase protein (T311). Extract of pigmented melanoma metastases from lymph nodes (HMB45)
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          200 µg/mL
          Amount
          20 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4, with 0.05% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          4°C, do not freeze
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          MART1 is involved in melanosome biogenesis by ensuring the stability of GPR143. Plays a vital role in the expression, stability, trafficking, and processing of melanocyte protein PMEL, which is critical to the formation of stage II melanosomes.
          Bioinformatics
          Protein Aliases: Antigen LB39-AA; Antigen SK29-AA; epididymis secretory sperm binding protein; MART-1; melanoma antigen recognized by T-cells; Melanoma antigen recognized by T-cells 1; MLANA; Protein Melan-A; unnamed protein product
          View more View less
          Gene Aliases: MART-1; MART1; MLANA
          View more View less
          UniProt ID: (Human) Q16655
          View more View less
          Entrez Gene ID: (Human) 2315
          View more View less
          Function(s)
          protein binding
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline