          • Primary Antibodies ›
          • HPV Type 16 L1 Antibodies
          NeoBiotechnologies
          HPV-16 (Human Papilloma Virus 16) Monoclonal Antibody (HPV16L1/1058), CF® 488
          View all (25) HPV Type 16 L1 antibodies
          Tech Support
          Cite HPV-16 (Human Papilloma Virus 16) Monoclonal Antibody (HPV16L1/1058), CF® 488
          Additional Information:
          {{banner.description}}
          Product Details
          MSM2-1058-CF488-100T
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:100
          -
          Flow Cytometry (Flow)
          5 µL/10^6 cells
          -
          Product Specifications
          Species Reactivity
          Virus
          Host/Isotype
          Mouse / IgG2a, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          HPV16L1/1058
          Immunogen
          Human papilloma virus type 16, major capsid protein L1
          Conjugate
          CF® 488 CF® 488 CF® 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          488/515 nm
          Form
          Liquid
          Concentration
          100 µg/mL
          Amount
          50 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4, with 0.05% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          L1 protein is a major capsid protein of HPV type 16. HPV 6 and 11 have been associated with relatively benign diseases such as genital warts but type 16 and 18 are strongly associated with cervical, vaginal and vulvar malignancies.
          Bioinformatics
          Protein Aliases: HPV L1; HPV-16; HPV16; human papillomavirus
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline